Skip to Content
Merck
All Photos(1)

Key Documents

218696

Sigma-Aldrich

Casein Kinase I Inhibitor, D4476

The Casein Kinase I Inhibitor, D4476, also referenced under CAS 301836-43-1, controls the biological activity of Casein Kinase I. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

Casein Kinase I Inhibitor, D4476, CK1 Inhibitor, 4-(4-(2,3-Dihydrobenzo[1,4]dioxin-6-yl)-5-pyridin-2-yl-1H-imidazol-2-yl)benzamide

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€223.00

€223.00


Estimated to ship on12 April 2025



Select a Size

Change View
1 MG
€223.00

About This Item

Empirical Formula (Hill Notation):
C23H18N4O3
CAS Number:
Molecular Weight:
398.41
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€223.00


Estimated to ship on12 April 2025


Quality Level

Assay

≥95% (HPLC)

form

crystalline solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

solubility

DMSO: 40 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

[nH]1c(nc(c1c5ncccc5)c3cc4c(cc3)OCCO4)c2ccc(cc2)C(=O)N

InChI

1S/C23H18N4O3/c24-22(28)14-4-6-15(7-5-14)23-26-20(21(27-23)17-3-1-2-10-25-17)16-8-9-18-19(13-16)30-12-11-29-18/h1-10,13H,11-12H2,(H2,24,28)(H,26,27)

InChI key

DPDZHVCKYBCJHW-UHFFFAOYSA-N

General description

A cell-permeable triaryl substituted imidazolo compound that acts as a potent, reversible and relatively specific ATP-competitive inhibitor of CK1 (IC50 = 0.2 µM from S. pombe; 0.3 µM for CK1δ) and ALK5 (IC50 = 0.5 µM). A weak inhibitor of p38α MAP kinase (IC50 = 12 µM) and only weakly affects the activities of a panel of kinases tested, including PKB, SGK and GSK-3β. Suppresses the site-specific phosphorylation (Ser322 and Ser325) and nuclear exclusion of FOXO1a in H4IIE hepatoma cells. Shown to be ~10-fold more potent than IC261 (Cat. No. 400090; IC50 = 2.5 µM for CK1).
A cell-permeable, reversible, potent, and relatively specific ATP-competitive inhibitor of CK1 (IC50 = 200 nM from S. pombe; 0.3 µM for CK1δ) and ALK5 (IC50 = 500 nM). A weak inhibitor of p38α MAP kinase (IC50 = 12 µM) and several other kinases tested, including PKB, SGK, and GSK-3β. Suppresses the site-specific phosphorylation (Ser322 and Ser325) and nuclear exclusion of FOXO1A in H4IIE hepatoma cells. Shown to be ~ 10-fold more potent than IC261 (Cat. No. 400090; IC50 = 2.5 µM for CK1).

Biochem/physiol Actions

Cell permeable: yes
Product competes with ATP.
Reversible: yes
Target IC50: 0.2 µM against CK1 from S. pombe; 0.3 µM for CK1δ 0.5 µM against ALK5

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael Ranes et al.
Molecular cell, 81(16), 3246-3261 (2021-08-06)
The Wnt/β-catenin pathway is a highly conserved, frequently mutated developmental and cancer pathway. Its output is defined mainly by β-catenin's phosphorylation- and ubiquitylation-dependent proteasomal degradation, initiated by the multi-protein β-catenin destruction complex. The precise mechanisms underlying destruction complex function have
Tianzhi Huang et al.
Molecular cell, 81(6), 1276-1291 (2021-02-05)
Aberrant cell proliferation is a hallmark of cancer, including glioblastoma (GBM). Here we report that protein arginine methyltransferase (PRMT) 6 activity is required for the proliferation, stem-like properties, and tumorigenicity of glioblastoma stem cells (GSCs), a subpopulation in GBM critical

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service