Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Pack Size | SKU | Availability | Price |
|---|
About This Item
UNSPSC Code:
41116012
NACRES:
NA.41
eCl@ss:
32160405
Pricing and availability is not currently available.
Application
Research Category
All
All
Physical form
1M Tris, 3M NaCl, pH 7.4.
Preparation Note
1 year at 4°C
Legal Information
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
1 of 1
This Item | |||
|---|---|---|---|
| form liquid | form liquid | form liquid | form - |
| manufacturer/tradename Upstate® | manufacturer/tradename Upstate® | manufacturer/tradename Chemicon®, IHC Select | manufacturer/tradename Calbiochem® |
| Quality Level 100 | Quality Level 100 | Quality Level 300 | Quality Level 100 |
| shipped in wet ice | shipped in wet ice | shipped in - | shipped in ambient |
Storage Class
10 - Combustible liquids
wgk
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
J Semba et al.
European journal of pharmacology, 230(3), 321-326 (1993-01-19)
The temporal inter-relationship of various amino acids and monoamine metabolites in rat cerebrospinal fluid was examined after acute administration of milacemide (100, 200 or 400 mg/kg i.p.), a glycine prodrug. Glycine concentrations rose linearly and dose dependently (20-190%) but were
D E Timm et al.
Protein science : a publication of the Protein Society, 3(3), 451-458 (1994-03-01)
Interactions between the purified recombinant receptor extracellular domain (RED) of the human low-affinity neurotrophin receptor (LANR) and recombinant human brain-derived neurotrophic factor, neurotrophin-3 (NT-3) and neuotrophin-4/5 have been studied by chemical crosslinking and circular dichroism. Conformational changes subsequent to binding
Chromatographic purification of the chloroplast ATP synthase (CF0-CF1) and the role of CF0 subunit IV in proton conduction.
Y Feng,R E McCarty
The Journal of Biological Chemistry null
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 20-190 | 04053252472053 |
| SYNC150190-1EA | 04061832190297 |
| BAF378711000-20EA | 04065269239369 |
| GF11136080-10EA | 04061825031132 |
| GF11136080-5EA | 04061825031163 |
| GF11136080-2EA | 04061825031156 |
| GF11136080-1EA | 04061825031149 |



