Skip to Content
Merck

Skip To

20-168

Mg2+ Lysis/Wash Buffer, 5X

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

UNSPSC Code:
41116012
NACRES:
NA.41
eCl@ss:
32160405
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

liquid

Quality Level

manufacturer/tradename

Upstate®

technique(s)

affinity binding assay: suitable (G-protein)

shipped in

dry ice

Application

Mg2+ Lysis/Wash Buffer, 5X may be used in G-protein assay. It is also suitable for use in activation assay kits.
Mg2+ Lysis/Wash Buffer, 5X may be used in G-protein assay. It is also suitable for use in activation assay kits.

Physical form

125mM HEPES, pH 7.5, 750mM NaCl, 5% Igepal CA-630, 50mM MgCl2, 5mM EDTA and 10% glycerol.

Preparation Note

1 year at -20°C

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
20-14820-14520-175
technique(s)

affinity binding assay: suitable (G-protein)

technique(s)

activity assay: suitable (histone acetyltransferase (HAT))

technique(s)

activity assay: suitable (kinase)

technique(s)

activity assay: suitable (histone deacetylase (HDAC))

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

form

liquid

form

liquid

form

liquid

form

liquid

manufacturer/tradename

Upstate®

manufacturer/tradename

Upstate®

manufacturer/tradename

Upstate®

manufacturer/tradename

Upstate®

shipped in

dry ice

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice


pictograms

CorrosionEnvironment

signalword

Danger

Storage Class

10 - Combustible liquids

wgk

WGK 3

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - ED ENV 1 - Eye Dam. 1


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

9036-19-5

CAS No.

9036-19-5

CAS No.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Vimal K Balasubramanian et al.
Current protocols, 1(5), e153-e153 (2021-05-28)
Plant organs and tissues contain multiple cell types, which are well organized in 3-dimensional structure to efficiently perform physiological functions such as homeostasis and response to environmental perturbation and pathogen infection. It is critically important to perform molecular measurements at
Mihály Cserepes et al.
Cancer research, 80(4), 663-674 (2020-01-01)
Clinical evidence shows that following initial response to treatment, drug-resistant cancer cells frequently evolve and, eventually, most tumors become resistant to all available therapies. We compiled a focused library consisting of >500 commercially available or newly synthetized 8-hydroxyquinoline (8OHQ) derivatives



Global Trade Item Number

SKUGTIN
20-16804053252738517
100801000104027269213848

Questions

Reviews

No rating value

Active Filters