Skip to Content
Merck

Skip To

20-105

Malachite Green Solution A

Synonym(s):

Malachite Green Dye, Malachite Green Reagent, Malachite Green Solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

UNSPSC Code:
41116012
NACRES:
NA.41
eCl@ss:
32160405
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

liquid

Quality Level

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (phosphatase)

shipped in

wet ice

Application

For use in the malachite green assay kit.
Research Category
All

Physical form

10mM ammonium molybdate, 1N HCl and 3.4% ethanol.

Preparation Note

2 years at 4°C

Analysis Note

Routinely evaluated by Malachite Green Assay.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
20-1041.106301.10329
technique(s)

activity assay: suitable (phosphatase)

technique(s)

activity assay: suitable (phosphatase)

technique(s)

-

technique(s)

microbiological culture: suitable

form

liquid

form

liquid

form

liquid

form

powder (dehydrated (DCM))

Quality Level

100

Quality Level

100

Quality Level

400

Quality Level

200

shipped in

wet ice

shipped in

wet ice

shipped in

-

shipped in

-

manufacturer/tradename

Upstate®

manufacturer/tradename

Upstate®

manufacturer/tradename

-

manufacturer/tradename

GranuCult®


pictograms

Corrosion

signalword

Warning

hcodes

Hazard Classifications

Aquatic Chronic 3 - Met. Corr. 1

flash_point_f

Not applicable

flash_point_c

Not applicable

Storage Class

12 - Non Combustible Liquids


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

12027-67-7

CAS No.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Isabelle Guisle et al.
Sleep, 43(4) (2019-11-09)
Aggregates of hyperphosphorylated tau protein are a hallmark of Alzheimer's disease (AD) and other tauopathies. Sleep disturbances are common in AD patients, and insufficient sleep may be a risk factor for AD. Recent evidence suggests that tau phosphorylation is dysregulated



Global Trade Item Number

SKUGTIN
20-10504053252286254
54020-U04061832566832

Questions

Reviews

No rating value

Active Filters