Skip to Content
Merck

Skip To

178221

α2-Antiplasmin, Human Plasma

A 452-amino acid glycoprotein present in plasma at about 70 µg/ml.

Synonym(s):

α2-Protease Inhibitor, Primary Plasmin Inhibitor

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
100 μg
Check Cart for Availability
€ 87,70

About This Item

NACRES:
NA.77
UNSPSC Code:
12352202
Assay:
≥95% (SDS-PAGE)
Form:
solid
Quality level:
Storage condition:
OK to freeze

€ 87,70


Check Cart for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Level

assay

≥95% (SDS-PAGE)

form

solid

specific activity

≥5 IU/mg protein

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

shipped in

ambient

storage temp.

−20°C

General description

A 452-amino acid glycoprotein present in plasma at about 70 µg/ml. Efficiently inhibits the plasminogen activator-induced lysis of fibrin clots. Forms a covalent complex with plasmin and inactivates it.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Plasminogen activator-induced lysis of fibrin clots
Product does not compete with ATP.
Reversible: no

Physical form

Lyophilized from 200 mM NaCl, 20 mM Bis-Tris, pH 6.4.

Preparation Note

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.
Please refer to the vial label for the lot-specific reconstitution volume.
Prepared from plasma that has been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV.

Other Notes

One Inhibitory Unit (I.U.) is defined as the amount of enzyme that will inhibit 1 unit plasmin activity using tosyl-Gly-Pro-Lys-pNA as substrate at 25°C, pH 7.8
Schaefer, B.M., et al. 1996. Arch. Dermatol. Res. 288, 122.
Aoki, N., et al. 1980. Blood 55, 483.
Saito, H., et al. 1974. Circ. Res.34, 641.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.
Toxicity: Standard Handling (A)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
178196441251178251
form

solid

form

solid

form

lyophilized

form

solid

assay

≥95% (SDS-PAGE)

assay

≥95% (SDS-PAGE)

assay

≥95% (SDS-PAGE)

assay

≥95% (SDS-PAGE)

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage condition

OK to freeze

storage condition

OK to freeze

storage condition

OK to freeze

storage condition

OK to freeze


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library





Global Trade Item Number

SKUGTIN
178221-100UG04055977223101

Questions

Reviews

No rating value

Active Filters