Skip to Content
Merck
All Photos(1)

Key Documents

1.70391

Supelco

Thorium ICP standard

traceable to SRM from NIST Th(NO₃)₄ in HNO₃ 2-3% 10 mg/l Th Certipur®

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€288.00

€288.00


Estimated to ship on24 May 2025


Request a Bulk Order

Select a Size

Change View
100 ML
€288.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

€288.00


Estimated to ship on24 May 2025


Request a Bulk Order

grade

certified reference material

Quality Level

Agency

suitable for EPA 200.8

product line

Certipur®

concentration

10 mg/L Th in 2-3% HNO3

technique(s)

ICP: suitable

pH

1 (20 °C in H2O)

density

1.012 g/cm3 at 20 °C

application(s)

industrial qc
pharmaceutical

format

single component solution

storage temp.

15-25°C

General description

This certified reference material (CRM) is produced in accordance with ISO 17034 and characterized in accordance with ISO/IEC 17025. This CRM is traceable to SI unit kg and measured against primary material from a National Metrology Institute (NMI), e.g. NIST.

Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation.

Download your certificate at http://www.sigma-aldrich.com to view certified values, including uncertainty, date of expiry, and detailed information about trace impurities.

Application

Thorium ICP standard may be used as a calibration standard for preconcentration and separation of thorium in water samples by ICP-MS technique[1].

Analysis Note

10 mg/L Th in 2-3% nitric acid, prepared with high-purity Th(NO3)4 and HNO3

Other Notes

For a complete product listing of our Certipur® range of CRMs for ICP and AAS, technical information, and example certificates please visit our ICP & AAS standards website

Legal Information

CERTIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of uranium and thorium in complex samples using chromatographic separation, ICP-MS and spectrophotometric detection
M Rozmaric, et.al.
Talanta, 80, 352-362 (2009)
Determination of uranium and thorium in complex samples using chromatographic separation, ICP-MS and spectrophotometric detection
M Rozmaric
Talanta, 80, 352-362 (2009)

Related Content

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Questions

  1. Is this an exempt quantity of thorium, or is a radioactive material license required?

    1 answer
    1. No license is required. Product 170391 contains a small amount of Th(NO3)4 in HNO3. It is in liquid form, not solid form. It is only labeled as "corrosive to metal" as listed in the Safety Data Sheet.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service