Skip to Content
Merck
All Photos(1)

Key Documents

1.05621

Supelco

Sodium silicate solution

extra pure

Sign Into View Organizational & Contract Pricing

Select a Size

2.5 L
€92.20
40 KG
€512.00
250 KG
€1,990.00

€92.20


Estimated to ship on15 April 2025


Request a Bulk Order

Select a Size

Change View
2.5 L
€92.20
40 KG
€512.00
250 KG
€1,990.00

About This Item

UNSPSC Code:
12352308
NACRES:
NB.21

€92.20


Estimated to ship on15 April 2025


Request a Bulk Order

Quality Level

form

liquid

potency

>2000 mg/kg LD50, oral (Rat)

pH

11.0-11.5 (20 °C, 50 g/L in H2O)

bp

>100 °C/1013 hPa

density

1.35 g/cm3 at 20 °C

storage temp.

15-25°C

Application


  • The Study′s Chemical Interaction of the Sodium Silicate Solution with Extender Pigments to Investigate High Heat Resistance Silicate Coating.: This research focuses on the geopolymerization process between sodium silicate solution and extender pigments (ZnO, TiO2, Fe2O3, CaCO3, and Na2SiF6). The study utilizes Fourier transform infrared spectroscopy (FT-IR), thermal gravimetric analysis (TGA), and X-ray diffraction (XRD) to analyze the chemical interactions. Findings indicate enhanced thermal stability of the coatings, making them suitable for high-temperature applications (Le et al., 2021).

  • Preparation and properties of metakaolin-fumed-silica geopolymer modified with sodium silicate and potassium silicate activators.: This paper explores the effects of sodium silicate and potassium silicate activators on the properties of metakaolin-fumed-silica geopolymers. It provides insights into the structural and mechanical properties through a combination of experimental techniques, demonstrating improvements in material performance for construction applications (Jin et al., 2024).

  • Unravelling the stability of solvated silicate species in sodium silicate activator solution of different silica modulus by FTIR and NMR studies and their chemical reactivity on metakaolin and fly ash geopolymerisation.: This study investigates the stability and reactivity of solvated silicate species in sodium silicate solutions with varying silica modulus. Using FTIR and NMR spectroscopy, it elucidates the mechanisms of geopolymerisation with metakaolin and fly ash, highlighting the formation of stable geopolymer structures suitable for sustainable construction materials (Sharmila et al., 2024).

  • Formation of Geopolymers Using Sodium Silicate Solution and Aluminum Orthophosphate.: This paper examines the formation and structural properties of geopolymers activated with sodium silicate solutions of varying modules and aluminum orthophosphate. Using X-ray diffraction and NMR measurements, the study reveals the reaction kinetics and structural evolution of the geopolymers, highlighting their potential for high-performance materials (Partschefeld et al., 2020).

Analysis Note

Identity: passes test
Assay (acidimetric, Na₂O): 7.5 - 8.5 %
Assay (acidimetric, SiO₂): 25.5 - 28.5 %
Density (20 °C): 1.296 - 1.396 g/ml
Carbonate (as CO₂): passes test
Fe (Iron): ≤ 50 ppm
Heavy metals (as Pb): ≤ 50 ppm

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service