Skip to Content
Merck
All Photos(2)

Key Documents

1468220080

Millipore

Tryptic Soy Agar - Ready-to-use Settle Plates

IsoBag®, DPTE Beta Bag with 190 mm Alpha Port, ICR plus lockable plate, sterile; γ-irradiated, plate diam. 90 mm, double packed of, suitable for air monitoring

Synonym(s):

CASO Agar, IsoBag® Tryptic Soy Agar Settle Plate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103403
NACRES:
NA.74
Pricing and availability is not currently available.

material

polystyrene plate

Quality Level

Agency

EP
USP

sterility

sterile; γ-irradiated

form

plate

feature

ready-to-use (Culture Media)

packaging

pack of 1 unit
double packed of

manufacturer/tradename

IsoBag®

technique(s)

microbiological culture: suitable

plate diam.

90 mm

pH

7.1-7.5

application(s)

air monitoring
pharmaceutical

storage temp.

15-25°C

suitability

bacteria
nonselective for

Looking for similar products? Visit Product Comparison Guide

General description

The IsoBag® TSA Settle Plus is designed for quick and convenient transfer of culture media for air monitoring into isolators or RABS systems. The IsoBag® TSA Settle Plus is a sterile polyurethane DPTE Beta Bag compatible with 190 mm DPTE Alpha Ports containing lockable 90 mm settle plates.  
The formulation of the Tryptic Soy Agar contains combination of casein and soy peptones that renders the medium nutritious by supplying organic nitrogen, particularly amino acids, and longer-chained peptides. Sodium chloride maintains the osmotic equilibrium and the medium is supplemented with pyruvate to provide an efficient neutralization of hydrogen peroxide.
The filled IsoBag® is double bagged, lockable, and irradiated in the final packaging to avoid cross contamination.

Application

IsoBag® Tryptic Soy Agar contact settle plates are designed to determine the total aerobic microbial count suitable for active and passive air monitoring inside isolators or RABS systems.

Legal Information

ISOBAG is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

IsoBag® ease the environmental monitoring workflow in aseptic production isolator

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service