Skip to Content
Merck
All Photos(1)

Key Documents

1.44242

Millipore

Hycon® Agar Strips YM

Rose-Bengal Chloramphenicol Agar, for yeasts, for molds, aseptically filled, single packed of 50 units, suitable for air monitoring

Synonym(s):

Hycon Ready-to-use Agar Strips, Rose-Bengal Agar Strips

Sign Into View Organizational & Contract Pricing

Select a Size

50 UNITS
€184.00

€184.00


Estimated to ship on29 May 2025


Request more information

Select a Size

Change View
50 UNITS
€184.00

About This Item

UNSPSC Code:
41113035
NACRES:
NA.74

€184.00


Estimated to ship on29 May 2025


Request more information

Quality Level

sterility

aseptically filled

product line

RCS®

form

plate (ready-to-use)

feature

individually wrapped

packaging

single packed of 50 units

manufacturer/tradename

HYCON®

pH

7.3 ( in H2O)

application(s)

air monitoring
cosmetics
food and beverages
pharmaceutical

compatibility

for use with RCS® High Flow Touch

storage temp.

2-8°C

suitability

molds
yeasts

Looking for similar products? Visit Product Comparison Guide

General description

Agar Strips YM are ready-to-use culture media for assessment of airborne yeasts and molds with RCS® Air Samplers. Each individual agar strip is welded into a re-sealable sleeve and 100% visually controlled, ensuring maximum safety in critical environments. Rose-Bengal chloramphenicol agar is filled in a flexible backing film. Rose-Bengal chloramphenicol agar is a selective medium for the enumeration of yeasts and molds. Chloramphenicol, streptomycin and Rose-Bengal are included as selective agents to inhibit bacterial growth. Rose-Bengal also restricts the size and height of the colonies of rapidly growing molds, which facilitates the detection of slow growing fungi.

Application

Agar Strips are used for monitoring of ambient air and compressed gases with RCS® Air Samplers.

Legal Information

HYCON is a registered trademark of Merck KGaA, Darmstadt, Germany
RCS is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service