Skip to Content
Merck
All Photos(1)

Key Documents

1.44228

Millipore

Hycon® Agar Strips TCI-y

TCI-y medium, Polysorbate 80, Thioglycollate, Lecithin, for bacteria, for fungi, γ-irradiated, double packed of 40 units, suitable for air monitoring

Synonym(s):

Hycon Ready-to-use Agar Strips, TSA, Soybean Casein Digest, Agar Strips

Sign Into View Organizational & Contract Pricing

Select a Size

40 UNITS
€184.00

€184.00


Estimated to ship on24 May 2025


Request more information

Select a Size

Change View
40 UNITS
€184.00

About This Item

UNSPSC Code:
41113035
NACRES:
NA.74

€184.00


Estimated to ship on24 May 2025


Request more information

Agency

EP

Quality Level

reg. compliance

FDA

sterility

γ-irradiated

form

plate (ready-to-use)

feature

individually wrapped

composition

Lecithin
Polysorbate 80
Thioglycollate

packaging

double packed of 40 units

manufacturer/tradename

HYCON®

pH

7.3 ( in H2O)

mp

>95 °C

application(s)

air monitoring
pharmaceutical

compatibility

for use with RCS® High Flow Touch

storage temp.

2-25°C

suitability

bacteria
fungi

Looking for similar products? Visit Product Comparison Guide

General description

Agar Strips TCI-y are ready-to-use culture media for assessment of airborne microorganisms with RCS® Air Samplers. Each individual agar strip is welded into a re-sealable sleeve and 100% visually controlled, ensuring maximum safety in critical environments. The basic medium is filled in a flexible backing film. TCI-y is a complex medium for cultivation and isolation of a wide range of bacteria, yeasts and molds. The medium is supplemented with neutralizers like lecithin, polysorbate 80 and thioglycollate. Addition of Pyruvate inactivates VHP residues, when used in isolators.

Application

Agar Strips are used for monitoring of ambient air and compressed gases with RCS® Air Samplers. These Agar strips gamma irradiated and triple bagged, for specific application within isolator and critical environment

Legal Information

HYCON is a registered trademark of Merck KGaA, Darmstadt, Germany
RCS is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

For easy and flexible microbiological monitoring of surfaces in the food & beverage industry.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service