Skip to Content
Merck
All Photos(1)

Key Documents

1.44240

Millipore

Hycon® Agar Strips TSM

Tryptic Soy Agar, Lecithin, Polysorbate 80, aseptically filled, single packed of 50 units, suitable for air monitoring

Synonym(s):

Hycon Ready-to-use Agar Strips, TSA, Soybean Casein Digest, Agar Strips

Sign Into View Organizational & Contract Pricing

Select a Size

50 UNITS
€184.00

€184.00


Estimated to ship on26 May 2025


Request more information

Select a Size

Change View
50 UNITS
€184.00

About This Item

UNSPSC Code:
41113035
NACRES:
NA.74

€184.00


Estimated to ship on26 May 2025


Request more information

Agency

EP
JP
USP

Quality Level

sterility

aseptically filled

form

plate (ready-to-use)

feature

individually wrapped

composition

Lecithin
Polysorbate 80

packaging

single packed of 50 units

manufacturer/tradename

HYCON®

pH

7.3 ( in H2O)

application(s)

air monitoring
cosmetics
food and beverages
pharmaceutical

compatibility

for use with RCS® High Flow Touch

storage temp.

2-25°C

suitability

bacteria
fungi

Looking for similar products? Visit Product Comparison Guide

General description

Agar Strips TSM are ready-to-use culture media for assessment of airborne microorganisms with RCS® Air Samplers. Each individual agar strip is welded into a re-sealable sleeve and 100% visually controlled, ensuring maximum safety in critical environments. Tryptic Soy Agar is filled in a flexible backing film. Tryptic Soy Agar (TSA, Tryptic Soy Agar Soybean Casein Digest Agar) is a complex medium for cultivation and isolation of a wide range of bacteria, yeasts and molds. The medium is supplemented with the neutralizers such as lecithin and polysorbate 80.

Application

Agar Strips are used for monitoring of ambient air and compressed gases with RCS® Air Samplers.

Legal Information

HYCON is a registered trademark of Merck KGaA, Darmstadt, Germany
RCS is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service