Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Pack Size | SKU | Availability | Price |
|---|---|---|---|
| 6 μg | Check Cart for Availability | € 554,00 |
About This Item
UNSPSC Code:
12352202
NACRES:
NA.32
eCl@ss:
32160405
Biological source:
human
Mol wt:
Mw 48 kDa
biological source
human
mol wt
Mw 48 kDa
manufacturer/tradename
Upstate®
technique(s)
activity assay: suitable (kinase)
NCBI accession no.
UniProt accession no.
Gene Information
human ... SGK1(6446)
General description
N-truncated (missing Nterminal 60 amino acids), His-tagged, human SGK1 fusion protein, containing a S422D mutation
Product Source: expressed in Sf21 cells
Biochem/physiol Actions
Protein Target: SGK1
Target Sub-Family: AGC
Physical form
Ni-NTA agarose
Preparation Note
1 year at -70°C
Analysis Note
routinely evaluated by Kinase assay
Other Notes
For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.
Legal Information
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
1 of 1
This Item | |||
|---|---|---|---|
| technique(s) activity assay: suitable (kinase) | technique(s) activity assay: suitable (kinase) | technique(s) activity assay: suitable (kinase) | technique(s) activity assay: suitable (kinase) |
| biological source human | biological source human | biological source human | biological source human |
| mol wt Mw 48 kDa | mol wt Mw 43 kDa | mol wt Mw 81.6 kDa | mol wt Mw 49.2 kDa |
| Gene Information human ... SGK1(6446) | Gene Information human ... AKT3(10000) | Gene Information human ... CHEK1(1111) | Gene Information human ... MAPK9(5601) |
| UniProt accession no. | UniProt accession no. | UniProt accession no. | UniProt accession no. |
| NCBI accession no. | NCBI accession no. | NCBI accession no. | NCBI accession no. |
signalword
Warning
hcodes
Hazard Classifications
Skin Sens. 1
Storage Class
10 - Combustible liquids
wgk
WGK 2
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Activation of NHE3 by dexamethasone requires phosphorylation of NHE3 at Ser663 by SGK1
Wang, Dongsheng, et al
American Journal of Physiology. Cell Physiology, 289, C802-C810 (2005)
Dana Yoo et al.
The Journal of biological chemistry, 278(25), 23066-23075 (2003-04-10)
The Kir1.1 (ROMK) subtypes of inward rectifier K+ channels mediate potassium secretion and regulate sodium chloride reabsorption in the kidney. The density of ROMK channels on the cortical collecting duct apical membrane is exquisitely regulated in concert with physiological demands.
T Kobayashi et al.
The Biochemical journal, 344 Pt 1, 189-197 (1999-11-05)
The catalytic domain of serum- and glucocorticoid-induced protein kinase (SGK) is 54% identical with protein kinase B (PKB) and, like PKB, is activated in vitro by 3-phosphoinositide-dependent protein kinase-1 (PDK1) and in vivo in response to signals that activate phosphatidylinositol
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 14-331 | 04053252328633 |




