1.25136
Fractogel® EMD TMAE (M)
RoboColumn Fractogel® TMAE (M), volume 0.6 mL, prepacked column
Synonym(s):
RoboColumn Fractogel® TMAE (M), 0.6 ml
Select a Size
€343.00
Estimated to ship on21 May 2025
Select a Size
About This Item
€343.00
Estimated to ship on21 May 2025
Recommended Products
ligand
(Trimethylammoniumethyl)
material
polypropylene housing
product line
Fractogel®
form
prepacked column
parameter
≤8 bar pressure
~240 cm/hr max. flow rate
column I.D. × L
5 mm × 30 mm
volume
0.6 mL
matrix active group
Methacrylate
mean particle size
40-90 μm
capacity
~100 mg, BSA binding capacity
application(s)
gene therapy
mAb
recombinant protein
separation technique
anion exchange
storage temp.
15-25°C
General description
Features and Benefits
- High protein loading conditions resulting in good selectivity and resolution with the tentacle technology
- Excellent binding to large viruses at increased loading
- Homogenous binding with high selectivity and purity
- Reduce operating costs and resin replacement with enduring durability - allowing for multiple cycles of column regeneration and sanitization
- Compatibility with 2.5 % (v/v) aqueous benzyl alcohol containing 150 mM NaCl storage solution
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service