Skip to Content
Merck
All Photos(1)

Key Documents

1.05242

Supelco

Dowex 1-X8 Ion Exchange Resin

60-170 mesh

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
€427.00

€427.00


Estimated to ship on02 April 2025


Request a Bulk Order

Select a Size

Change View
250 ML
€427.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
41115711
NACRES:
NB.21

€427.00


Estimated to ship on02 April 2025


Request a Bulk Order

Product Name

Ion exchanger Dowex 1-X8, suitable for ion chromatography

form

resin

Quality Level

technique(s)

HPLC: suitable
LPLC: suitable
ion chromatography: suitable (strongly basic)

matrix

PS-DVB

matrix active group

SAX

particle size

90-250 μm

separation technique

anion exchange

storage temp.

15-25°C

SMILES string

[Cl-].[N+](C)(C)(C)c3ccc(cc3)C=C.c2(ccc(cc2)C=C)C=C.c1(ccccc1)C=C

InChI key

BBQMUEOYPPPODD-UHFFFAOYSA-M

General description

(strongly basic anion exchanger), Cl⁻ form

Application


  • Preconcentration of molybdenum, antimony and vanadium in gasoline samples using Dowex 1-x8 resin and their determination with inductively coupled plasma-optical emission spectrometry.: This study demonstrates the efficiency of Dowex 1-X8 ion-exchange resin in the preconcentration of trace metals from complex matrices, facilitating their quantification via ICP-OES, a technique pivotal for ensuring fuel quality and compliance with environmental regulations (Nomngongo et al., 2013).

  • Anion-exchange chromatographic separation of Hg for isotope ratio measurements by multicollector ICPMS.: Explores the use of Dowex 1-X8 for the chromatographic separation of mercury isotopes, enhancing the precision of isotope ratio analysis which is crucial in geochemical and environmental studies (Malinovsky et al., 2008).

  • Uranium removal from contaminated groundwater by synthetic resins.: Discusses the application of Dowex 1-X8 in the removal of uranium from water sources, highlighting its potential in remediation efforts at sites affected by radioactive contamination (Phillips et al., 2008).

  • Selective colorimetric determination of TNT partitioned between an alkaline solution and a strongly basic Dowex 1-X8 anion exchanger.: Presents a novel approach for the detection of trinitrotoluene (TNT) using Dowex 1-X8, which could be employed in forensic science to improve the sensitivity and selectivity of explosive residue analysis (Uzer et al., 2008).

  • Fluidized beds in flow analysis: use with ion-exchange separation for spectrophotometric determination of zinc in plant digests.: Showcases the integration of Dowex 1-X8 in flow analysis systems for the efficient separation and subsequent spectrophotometric determination of zinc, essential for agricultural and nutritional research (Ribeiro et al., 2006).

Analysis Note

Description: pale yellow moist spherules
Total capacity (wet): ≥ 1.2 mol/l
Particle size (0,09-0,25 mm): ≥ 70 %

Legal Information

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service