Skip to Content
Merck
All Photos(1)

Key Documents

1.25147

Millipore

Fractogel® EMD SE Hicap (M)

RoboColumn Fractogel® SE HC, volume 0.2 mL, prepacked column

Synonym(s):

RoboColumn Fractogel® SE HC 0.2ml

Sign Into View Organizational & Contract Pricing

Select a Size

8 PKG
€291.00

€291.00


Estimated to ship on29 May 2025



Select a Size

Change View
8 PKG
€291.00

About This Item

UNSPSC Code:
41115711

€291.00


Estimated to ship on29 May 2025


ligand

(Sulfoethyl)

Quality Level

material

polypropylene housing

product line

Fractogel®

form

prepacked column

parameter

≤8 bar pressure
~220 cm/hr max. flow rate

column I.D. × L

5 mm × 10 mm

volume

0.2 mL

matrix active group

Methacrylate

mean particle size

40-90 μm

capacity

~160 mL, resin binding capacity

application(s)

mAb
recombinant protein
vaccine development

separation technique

cation exchange

storage temp.

no temp limit

General description

5 x 10mm

Features and Benefits

RoboColumn Fractogel® SE HiCap (M) 0.2 ml features include:

  • Tentacle technology provides high yield, selectivity, and focused elution
  • High loading capacity combined with high purity at lower elution volumes
  • Compatibility with 2.5 % (v/v) aqueous benzyl alcohol containing 150 mM NaCl storage solution

Legal Information

FRACTOGEL is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service