Skip to Content
Merck

Skip To

12-434

Woodtide

Sign In to View Organizational & Contract Pricing.

About This Item

UNSPSC Code:
12352202
NACRES:
NA.41
eCl@ss:
32160405


manufacturer/tradename

Upstate®

Quality Segment

technique(s)

activity assay: suitable (kinase)

UniProt accession no.

shipped in

wet ice

Biochem/physiol Actions

Protein Target: DYRK1A

Physical form

sterile water

Preparation Note

1 year at -20°C

Analysis Note

Routinely evaluated as a substrate for DYRK1A and DYRK2.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
12-54512-493SRP0684
technique(s)

activity assay: suitable (kinase)

technique(s)

activity assay: suitable (kinase)

technique(s)

activity assay: suitable (kinase)

technique(s)

-

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

-

UniProt accession no.

Q13627

UniProt accession no.

O43293

UniProt accession no.

P00519

UniProt accession no.

-

shipped in

wet ice

shipped in

dry ice

shipped in

dry ice

shipped in

ambient

manufacturer/tradename

Upstate®

manufacturer/tradename

Upstate®

manufacturer/tradename

Upstate®

manufacturer/tradename

-


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library







Global Trade Item Number

SKUGTIN
12-43404053252272868

Questions

Reviews

No rating value

Active Filters