Skip to Content
Merck

Skip To

124013

Akt Inhibitor VI, Akt-in

The Akt Inhibitor VI, Akt-in controls the biological activity of Akt. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

Akt Inhibitor VI, Akt-in, TCL110-24, H-AVTDHPDRLWAWEKF-OH

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
2 mg
Check Cart for Availability
€ 262,00

About This Item

Empirical Formula (Hill Notation):
C88H123N23O23
Molecular Weight:
1871.06
NACRES:
NA.77
UNSPSC Code:
12352200
Assay:
≥95% (HPLC)
Form:
lyophilized solid
Storage condition:
OK to freeze, desiccated (hygroscopic)

€ 262,00


Check Cart for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥95% (HPLC)

form

lyophilized solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, desiccated (hygroscopic)

color

white

solubility

water: 1 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

A reversible 15-mer peptide derived from proto-oncogene TCL1 (amino acids 10-24) that acts as a specific inhibitor of Akt. Shown to bind to Akt-PH domain (Kd ~18 µM) and interfere with the Akt-phosphoinositide interaction, thus hindering membrane translocation from the cytosol and downstream biological responses.
A reversible 15-mer peptide derived from the amino acids 10-24 of the proto-oncogene TCL1 (TCL10-24) and acts as a specific inhibitor of Akt. Shown to bind to the Akt-PH domain (Kd ~18 µM) and interfere with the Akt-phosphoinositide interaction. This peptide, when made cell-permeable (Cat. No. 124014), has been shown to hinder Akt membrane translocation from the cytosol and subsequent downstream biological responses.

Biochem/physiol Actions

Cell permeable: no
Kd ~18 µM in binding to Akt-PH domain
Primary Target
Akt
Product does not compete with ATP.
Reversible: yes

Packaging

Packaged under inert gas

Physical form

Supplied as an acetate salt.

Preparation Note

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

H-Ala-Val-Thr-Asp-His-Pro-Asp-Arg-Leu-Trp-Ala-Trp-Glu-Lys-Phe-OH
Hiromura, M., et al. 2004. J. Biol. Chem.279, 53407.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Carcinogenic / Teratogenic (D)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
124014328000328005
assay

≥95% (HPLC)

assay

≥95% (HPLC)

assay

≥95% (HPLC)

assay

≥95% (HPLC)

form

lyophilized solid

form

solid

form

lyophilized solid

form

lyophilized solid

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

−20°C

solubility

water: 1 mg/mL

solubility

water: 1 mg/mL

solubility

water: 1 mg/mL, DMSO: 5 mg/mL

solubility

water: 1 mg/mL


Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content






Global Trade Item Number

SKUGTIN
124013-2MG04055977205657

Questions

Reviews

No rating value

Active Filters