Skip to Content
Merck

Skip To

12-243

S6 Kinase/Rsk2 Substrate Peptide 2

S6 Kinase/Rsk2 Substrate Peptide 2 primarily used in Kinase Assays.

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

UNSPSC Code:
12352202
NACRES:
NA.41
eCl@ss:
32160405
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

Gene Information

human ... RPS6KA3(6197)

Biochem/physiol Actions

Protein Target: Rsk2
Target Sub-Family: AGC

Physical form

Lyophilized powder

Preparation Note

Lyophilized: Stable for 2 years at -20°C

Analysis Note

Routinely evaluated as a substrate for p90 Rsk-2 and p70 S6 Kinase.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
12-42012-33020-122
technique(s)

activity assay: suitable (kinase)

technique(s)

activity assay: suitable (kinase)

technique(s)

activity assay: suitable (kinase)

technique(s)

activity assay: suitable (kinase)

Quality Level

100

Quality Level

100

Quality Level

300

Quality Level

100

UniProt accession no.

P51812

UniProt accession no.

Q15418

UniProt accession no.

P67870

UniProt accession no.

O75582, O75676, P23443, P51812, Q15349, Q15418, Q96S38, Q9UBS0, Q9UK32

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

Gene Information

human ... RPS6KA3(6197)

Gene Information

human ... RPS6KA1(6195)

Gene Information

human ... CSNK2B(1460)

Gene Information

human ... RPS6KA1(6195)

NCBI accession no.

NM_004586.2

NCBI accession no.

NM_001006665.1, NM_002953.3

NCBI accession no.

NM_001320.5

NCBI accession no.

NM_001006665.1, NM_002953.3


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library







Global Trade Item Number

SKUGTIN
12-24304053252338595

Questions

Reviews

No rating value

Active Filters