Skip to Content
Merck
All Photos(1)

Key Documents

1.18342

Supelco

Calibration Standard Set

turbidity 10000 NTU±200

Sign Into View Organizational & Contract Pricing

Select a Size

1 UNIT
€676.00

€676.00


Estimated to ship on29 May 2025


Request a Bulk OrderRequest more information

Select a Size

Change View
1 UNIT
€676.00

About This Item

UNSPSC Code:
47101516
NACRES:
NA.24

€676.00


Estimated to ship on29 May 2025


Request a Bulk OrderRequest more information

Product Name

Calibration standard 3000 IR, turbidity 10000 NTU±200

technique(s)

photometry: suitable

turbidity

10000 NTU±200

compatibility

for use with Turbiquant

storage temp.

15-25°C

General description

10 000 NTU Turbiquant
The Turbiquant Calibration Standard is suitable to calibrate the Turbiquant 3000 IR. The standard offers the following advantages:
1. The new primary standards have approvals which secondary standards do not have (rated to be a primary standard according to US EPA).
2. Instruments used in drinking water analysis no longer need to be certified with separate primary standards.
3. Clear records of final inspection are included in each set.
4. The new standards have an unrivalled precision due to production accuracy, and stability in solution.
5. The standards are ready-to-use.
6. They are non-toxic.
7. They are unproblematic to store and transport.

Analysis Note

Measured value (standard): 10000 FNU ±2 %

Legal Information

TURBIQUANT is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service