Skip to Content
Merck
All Photos(1)

Key Documents

1.18320

Supelco

Empty cells

Sign Into View Organizational & Contract Pricing

Select a Size

3 UNITS
€331.00

€331.00


Estimated to ship on29 May 2025


Request a Bulk OrderRequest more information

Select a Size

Change View
3 UNITS
€331.00

About This Item

UNSPSC Code:
47101516
NACRES:
SE.11

€331.00


Estimated to ship on29 May 2025


Request a Bulk OrderRequest more information

material

optical glass vial

technique(s)

photometry: suitable

compatibility

for use with Turbiquant 1000
for use with Turbiquant 1100

detection method

turbidimetric

storage temp.

2-30°C

General description

1000/1100 Turbiquant
These empty celles are suitable for the measurement of turbidity using the instrument types Turbiquant 1000 and 1100. The cells have a diameter of 24 mm, are equipped with screw caps, and are delivered in a seperate bowes to protect them from scratching.

Legal Information

TURBIQUANT is a trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service