Skip to Content
Merck
All Photos(1)

Key Documents

1.18381

Supelco

Calibration Standard Set

Sign Into View Organizational & Contract Pricing

Select a Size

1 UNIT
€173.00

€173.00


Estimated to ship on21 May 2025


Request a Bulk OrderRequest more information

Select a Size

Change View
1 UNIT
€173.00

About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

€173.00


Estimated to ship on21 May 2025


Request a Bulk OrderRequest more information

Product Name

Calibration standard,

form

liquid

technique(s)

photometry: suitable

compatibility

for use with Turbiquant 1500 IR
for use with Turbiquant 1500 T
for use with Turbiquant 3000 IR
for use with Turbiquant 3000 T

storage temp.

15-25°C

General description

1500 IR, 1500 T, 3000 IR, 3000 T 10.0 NTU Turbiquant
The Turbiquant Calibration Standard is suitable to calibrate the Turbiquant 1500 IR, 1500 T, 3000 IR, and 3000 T. The standard offers the following advantages:
1. The new primary standards have approvals which secondary standards do not have (rated to be a primary standard according to US EPA).
2. Instruments used in drinking water analysis no longer need to be certified with separate primary standards.
3. Clear records of final inspection are included in each set.
4. The new standards have an unrivalled precision due to production accuracy, and stability in solution.
5. The standards are ready-to-use.
6. They are non-toxic.
7. They are unproblematic to store and transport.

Legal Information

TURBIQUANT is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XIV (Authorisation List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service