Key Documents
1.17319
Millipore
RapidMicroBio Base Easy Clean
for use with MAS-100® Iso
Select a Size
1 EACH
€4,890.00
€4,890.00
Estimated to ship on30 May 2025
Request more information
Select a Size
Change View
1 EACH
€4,890.00
About This Item
UNSPSC Code:
41122409
NACRES:
NA.74
€4,890.00
Estimated to ship on30 May 2025
Request more information
material
stainless steel base
Agency
non-compendial
feature
installed
application(s)
air monitoring
pharmaceutical
compatibility
for use with MAS-100® Iso
General description
The RapidMicroBio Base Easy Clean, SST without perforated lid is designed for use with all variants of MAS-100 ISO NT® and MAS-100 ISO MH® remote microbial air samplers.
It is compatible with environmental monitoring cassettes of the Growth Direct® System, made from stainless steel and autoclavable.
This product is compatible with a specific RapidMicroBio Perforated Lid designed for Growth Direct® cassettes (article 1.17314).
Application
The RapidMicroBio Base Easy Clean, SST without perforated lid for MAS-100 ISO NT®/ MAS-100 ISO MH® remote microbial air samplers is used for viable air monitoring in isolators and RABS with Growth Direct® environmental monitoring cassettes.
Legal Information
Growth Direct is a registered trademark of Rapid Micro Biosystems, Inc.
MAS-100 is a registered trademark of MBV AG
MAS-100 ISO MH is a registered trademark of MBV AG
MAS-100 ISO NT is a registered trademark of MBV AG