Skip to Content
Merck

Skip To

1.16997

Total Hardness Test Strips, reflectometric

water hardness 0.1-30.0 °d, REFLECTOQUANT®, for use with RQflex® 20

Synonym(s):

Total Hardness chemical test strips, Total Hardness test strip for instrument reader

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
50 tests
Check Cart for Availability
€ 82,10

About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

€ 82,10


Check Cart for Availability

Request a Bulk OrderRequest more information
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Total Hardness Test, reflectometric, water hardness 0.1-30.0 °d, for use with REFLECTOQUANT®

description

quantitative

Quality Segment

specific analyte(s)

calcium, magnesium

water hardness measuring range

0.1-30.0 °d

application(s)

boiler and cooling water
drinking water
ground and surface water
mineral water

compatibility

RQflex® 20, for use with REFLECTOQUANT®

detection method

reflectometric

storage temp.

15-25°C

General description

Method: reflectometric with test strips 0.1 - 30.0 °d Reflectoquant®
The Reflectoquant total hardness test strips can be used to measure water hardness. The hardness (total hardness) is due to its content of salts of the alkaline earth metals calcium, magnesium, strontium, and barium (“hardening constituents”, later compounds only in traces). The reaction zones of the test strip, which are impregnated with differing amounts of Titriplex III, assume a green to red color, depending on the hardness of the water sample. The color is then determined reflectometrically on the RQflex® 20. For pre-testing a sample in an economic and quick way to estimate the hardness and a possible dilution factor prior to using this quantitative method we recommend our semi-quantitative MQuant test strips for total hardness. Further quantitative determination methods can be found with our Spectroquant tests.

Analysis Note

function control: conforms
Assay
- standard 1:
3.1 - 3.9 °d
- standard 2: 8.8 - 10.8 °d
- standard 3: 22.7 - 27.7 °d

Legal Information

REFLECTOQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
RQFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1.100251.100461.11104
detection method

reflectometric

detection method

colorimetric

detection method

colorimetric

detection method

titrimetric

specific analyte(s)

calcium, magnesium

specific analyte(s)

calcium, magnesium

specific analyte(s)

calcium, magnesium

specific analyte(s)

calcium, magnesium

compatibility

RQflex® 20, for use with REFLECTOQUANT®

compatibility

Reflectoquant®, Spectroquant®

compatibility

for use with MQuant® StripScan App

compatibility

for use with MQUANT®

storage temp.

15-25°C

storage temp.

15-25°C

storage temp.

15-25°C

storage temp.

15-25°C

application(s)

boiler and cooling water
drinking water
ground and surface water
mineral water

application(s)

drinking water
ground and surface water
mineral water

application(s)

drinking water
ground and surface water
mineral water

application(s)

-

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100


Storage Class

11 - Combustible Solids

wgk

WGK 1


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

288-32-4

CAS No.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library





Global Trade Item Number

SKUGTIN
116997000104022536169965

Questions

  1. How to test Hardness using these strips. Kindly guide. Do you provide any colour change chart to analyse indicative value of water Hardness in terms of Ca and Mg.

    1 answer
    1. These test strips are designed for use with the REFLECTOQUANT® RQflex® 20 reflectometer. Calcium and magnesium ions in a sample react with phthalein purple embedded in the reaction zone of the test strip to form a violet dye, which is measured quantitatively using the reflectometer. Each REFLECTOQUANT® test strip product includes a bar code scanned by the reflectometer to load a preset calibrated method. A color indication chart is included only with the colorimetric versions of the Total Hardness Test Strips.

      Helpful?

Reviews

No rating value

Active Filters