Skip to Content
Merck

1.14894

Millipore

Fractogel® EMD SE Hicap (M)

Synonym(s):

Fractogel® EMD SE Hicap (M)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115711
Pricing and availability is not currently available.

ligand

(Sulfoethyl)

Quality Level

description

strong cation exchanger, suspension in 20% ethanol and 150 mM NaCl (40-90 µm)

sterility

sterile (Caustic Stable)

product line

Fractogel®

form

resin

specific analyte(s)

MAbs

parameter

220 cm/hr flow rate
8 bar max. pressure

matrix active group

methacrylate

mean particle size

40-90 μm

capacity

160 mg binding capacity (lysozyme/ml of resin)

transition temp

flash point 35 °C (calculated)

application(s)

recombinant protein
vaccine development

separation technique

cation exchange
strong cation exchange

storage temp.

2-30°C

Looking for similar products? Visit Product Comparison Guide

General description

Strong cation exchange Fractogel® resin featuring the tentacle technology, for purification of basic and neutral proteins (e.g. antibodies) and peptides

Features and Benefits

Fractogel® EMD SE Hicap (M) features include:

  • Tentacle technology provides high yield, selectivity, and focused elution
  • High loading capacity combined with high purity at lower elution volumes
  • Compatibility with 2.5 % (v/v) aqueous benzyl alcohol containing 150 mM NaCl storage solution

Packaging

  • 1.14894.0100: Fractogel® EMD SE Hicap (M) Resin 100ml
  • 1.14894.0010: Fractogel® EMD SE Hicap (M) Resin 10ml
  • 1.14894.0500: Fractogel® EMD SE Hicap (M) Resin 500ml
  • 1.14894.5000: Fractogel® EMD SE Hicap (M) Resin 5L

Plastic bottle

Analysis Note

Appearance: Milky turbid suspension,free from impurities(foreign particles)
Microscopic evaluation: Uniform spherical particles,no agglomerates,no fines
Extractable matter (water): ≤ 0.08 %
Cerium: ≤ 20 µg/g
Pressure drop(column: ID=1.6 cm, L=10 cm at 5 ml/min): ≤ 1.0 bar
Particle size (d10): 37 - 45 µm
Particle size (d50): 48 - 60 µm
Particle size (d90): 63 - 77 µm
Colony forming units (TAMC + TYMC): ≤ 100 CFU/ml
Endotoxins: ≤ 1.00 EU/ml
Protein binding capacity (lysozyme): 120 - 160 mg/ml
Functional test (b:a): ≤ 0.20
Functional test: Separation of chymotrypsinogen A and lysozyme

Legal Information

FRACTOGEL is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

95.0 °F

Flash Point(C)

35 °C


What is the Emprove® Program?

The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials. Four document types are included with an Emprove® Program subscription:

Material Qualification Dossier

Free of charge - Sign in to download

Quality Management Dossier (Open Access)

Free of charge - Sign in to download

Operational Excellence Dossier

Available by subscription or for a fee

Learn more about the benefits an Emprove® Program subscription can provide
Why are these documents unavailable?
Emprove® Dossiers contain information intended to support the registration process and risk assessment activities of pharmaceutical producers and contain confidential information. Please contact us if you would like access.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service