Skip to Content
Merck

Skip To

1.02482

Cellvento® CHO-110

Chemically defined cell culture medium, dry powder

Sign In to View Organizational & Contract Pricing.

About This Item

UNSPSC Code:
41106213


Quality Segment

description

shipped with cooling packs

product line

Cellvento®

form

dry powder

technique(s)

cell culture | mammalian: suitable (suspension)

application(s)

pharma/biopharma processes

storage temp.

2-8°C

General description

Cellvento® CHO-110 chemically defined cell culture medium has been specially developed for the growth of Chinese Hamster Ovary (CHO) cells and the expression of monoclonal antibodies and recombinant proteins in suspension culture. The formulation is of non-animal origin, chemically defined and contains no hydrolysates or components of unknown composition. Cellvento® CHO-110 medium has been formulated without L-glutamine, hypoxanthine, and tymidine to keep flexibility in applications. It is available in dry powder form or as ready-to-use medium to fit to different experimental set-ups.

Application

Cellvento® CHO-110 medium:
  • has been designed to support optimized cell growth and performance with DHFR-negative CHO suspension cell types, primarily recombinant CHODG44 cell lines, but may be suitable for use with other CHO cell lines.
  • should be used for cell adaptation and cell bank generation.
  • is suitable for production in batch and perfusion culture applications.
  • is suitable for seed train expansion up to the N-2 step for fed-batch processes, switching to Cellvento® CHO210 medium at the final N-1 expansion step, which is then used in conjunction with Cellvento® Feed-210 companion feed in the final production culture.
This product is intended for research or further manufacturing but not for human or therapeutic use.

Preparation Note

Formulated to contain 27.5 grams of powder per liter of medium.

Legal Information

CELLVENTO is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1.018851.025771.01883
technique(s)

cell culture | mammalian: suitable (suspension)

technique(s)

cell culture | mammalian: suitable (suspension)

technique(s)

cell culture | mammalian: suitable (suspension)

technique(s)

cell culture | mammalian: suitable (suspension)

form

dry powder

form

dry powder

form

dry powder

form

dry powder

product line

Cellvento®

product line

Cellvento®

product line

Cellvento®

product line

Cellvento®

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

Quality Level

500

Quality Level

500

Quality Level

500

Quality Level

500

description

shipped with cooling packs

description

shipped with cooling packs

description

shipped with cooling packs

description

shipped with cooling packs


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Sens. 1

Storage Class

11 - Combustible Solids

wgk

WGK 2

hcodes



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters