1.14879
TOC Test, photometric
photometric, 50-800 mg/L (TOC), Spectroquant®
About This Item
Recommended Products
product name
TOC Cell Test, photometric, 50-800 mg/L (TOC), Spectroquant®
product line
Spectroquant®
Quality Level
usage
sufficient for 25 tests
analyte chemical class(es)
total organic carbon (TOC)
measuring range
50-800 mg/L (TOC)
technique(s)
photometry: suitable
compatibility
for use with Spectroquant® Nova 60 A
for use with Spectroquant® Prove 100
for use with Spectroquant® Prove 300
for use with Spectroquant® Prove 600
detection method
photometric (Indicator)
storage temp.
15-25°C
General description
This Spectroquant® TOC test allows the accurate quantification of the TOC especially in sewage inflow samples. The term TOC (Total Organic Carbon) is used to describe the total content of organically bound carbon in dissolved and undissolved forms. By digestion with sulfuric acid and peroxodisulfate, these compounds are transformed into carbon dioxide.
This reacts with an indicator, the color is determined photometrically. Inorganic carbon (dissolved carbon dioxide and carbonates) is expelled beforehand by acidification.
The digestion is analogous to APHA 5310 C (2014).
All our Cell and Reagent Test Kits are equipped with the unique Live ID (2D barcode) which allows seamless method recognition and contains essential information such as lot number, expiry date, and automatic calibration updates.
The Spectroquant®Cell Tests come with prefilled 16 mm round cells as well as all the required reagents to perform the analysis according to the procedure described in the accompanying instruction leaflet.
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2 - Met. Corr. 1 - Ox. Liq. 3 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
5.1B - Oxidizing hazardous materials
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service