Skip to Content
Merck
All Photos(1)

Key Documents

1.00857

Supelco

Silicate (Silicic Acid) Test, photometric

photometric, 1.1-1070 mg/L (SiO2), 0.5-500 mg/L (Si), Spectroquant®

Sign Into View Organizational & Contract Pricing

Select a Size

100 TESTS
€167.00

€167.00


Estimated to ship on29 May 2025


Request a Bulk OrderRequest more information

Select a Size

Change View
100 TESTS
€167.00

About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

€167.00


Estimated to ship on29 May 2025


Request a Bulk OrderRequest more information

Product Name

Silicate (Silicic Acid) Test, photometric, 1.1-1070 mg/L (SiO2), 0.5-500 mg/L (Si), Spectroquant®

product line

Spectroquant®

Quality Level

usage

sufficient for 100 tests

specific analyte(s)

silicate

measuring range

0.5-500 mg/L (Si)
1.1-1070 mg/L (SiO2)

technique(s)

photometry: suitable

compatibility

for use with Spectroquant® Move 100
for use with Spectroquant® Nova 60 A
for use with Spectroquant® Prove 100
for use with Spectroquant® Prove 300
for use with Spectroquant® Prove 600

detection method

photometric (Molybdosilicate)

storage temp.

15-25°C

General description

Method: photometric 1.1 - 1070 mg/l SiO₂ 0.5 - 500 mg/l Si Spectroquant®

The method is analogous to APHA 4500-SiO2 C.


This Spectroquant® Silicate Reagent Test allows the accurate quantification of silicate (silicic acid) in aqueous samples.


Method applied: In sulfuric solution silicate ions react with molybdate ions to form a yellow heteropoly acid which is determined photometrically.


The Spectroquant® Reagent Test Kits contain highly stable, ready-to-use reagent mixtures to perform the analysis according to the procedure described in the accompanying instruction leaflet .


All our Cell and Reagent Test Kits are equipped with the unique Live ID (2D barcode) which allows seamless method recognition and contains essential information such as lot number, expiry date, and automatic calibration updates.

Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service