Skip to Content
Merck
All Photos(1)

Documents

1.10130

Millipore

Potato dextrose agar

GranuCult® prime, USP, APHA, granular, suitable for pharmaceutical

Synonym(s):

PDA, Potato Glucose Agar

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106200
NACRES:
NA.74

Agency

APHA
USP

Quality Level

sterility

non-sterile

form

granular

manufacturer/tradename

GranuCult® prime

technique(s)

microbiological culture: suitable

pH

5.6 (21 °C, 39 g/L in H2O, after autoclaving)

solubility

39 g/L

bulk density

610 kg/m3

application(s)

microbiology
pharmaceutical

storage temp.

15-25°C

suitability

molds
yeasts

Linkage

Replaces: 1.10130.5007

Analysis Note

Appearance (clearness): clear to slightly opalescent
Appearance (color): yellowish-braun
pH-value (25 °C): 5.4 - 5.8
Growth (Geotrichum candidum DSM 1240): good to very good
Growth (Penicillium commune ATCC 10428): medium to good
Growth (Trichophyton ajelloi ATCC 28454): medium to good
Incubation: 5 days; 28 °C
Growth promotion test in accordance with the harmonised method of EP and USP:
Inoculum on reference medium (Candida albicans ATCC 10231 (WDCM 00054)): 10 - 100
Inoculum on reference medium (Aspergillus brasiliensis (formerly A. niger) ATCC 16404 (WDCM 00053)): 10 - 100
Inoculum on reference medium (Saccharomyces cerevisiae ATCC 9763 (WDCM 00058)): 10 - 100
Inoculum on reference medium (Rhodotorula mucilaginosa DSM 70403): 10 - 100
Colony count (Candida albicans ATCC 10231 (WDCM 00054)):
Colony count (Aspergillus brasiliensis (formerly A. niger) ATCC 16404 (WDCM 00053)):
Colony count (Saccharomyces cerevisiae ATCC 9763 (WDCM 00058)):
Colony count (Rhodotorula mucilaginosa DSM 70403):
Recovery on test medium (Candida albicans ATCC 10231 (WDCM 00054)): ≥ 70 %
Recovery on test medium (Aspergillus brasiliensis (formerly A. niger) ATCC 16404 (WDCM 00053)): ≥ 50 %
Recovery on test medium (Saccharomyces cerevisiae ATCC 9763 (WDCM 00058)): ≥ 70 %
Recovery on test medium (Rhodotorula mucilaginosa DSM 70403): ≥ 70 %
Incubation: up to 5 days; 20-25 °C

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Learn more about our culture media portfolio for microbial examination of non-sterile products, fully compliant with the harmonized Pharmacopoeia.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service