Skip to Content
Merck
All Photos(1)

Key Documents

1.14785

Supelco

Nickel Test, photometric

photometric, 0.02-5.00 mg/L (Ni), Spectroquant®

Sign Into View Organizational & Contract Pricing

Select a Size

250 TESTS
€213.00

€213.00


Estimated to ship on21 May 2025


Request a Bulk OrderRequest more information

Select a Size

Change View
250 TESTS
€213.00

About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

€213.00


Estimated to ship on21 May 2025


Request a Bulk OrderRequest more information

Product Name

Nickel Test, photometric, 0.02-5.00 mg/L (Ni), Spectroquant®

product line

Spectroquant®

Quality Level

usage

sufficient for 250 tests
250 tests

specific analyte(s)

nickel

measuring range

0.02-5.00 mg/L (Ni)

technique(s)

photometry: suitable

compatibility

for use with Spectroquant® Move 100
for use with Spectroquant® Nova 60 A
for use with Spectroquant® Prove 100
for use with Spectroquant® Prove 300
for use with Spectroquant® Prove 600

detection method

photometric (Dimethylglyoxime)

storage temp.

15-25°C

General description

Method: photometric 0.02 - 5.00 mg/l Ni Spectroquant®
This Spectroquant® Nickel test allows the accurate quantification of nickel ions in various waters. Nickel(II) ions are oxidized by iodine and then transformed with dimethylglyoxime in an ammoniacal solution into a red-brown complex that is determined photometrically.
MQuant® test strips can be used for pre-testing a sample in an economic and quick way to estimate the nickel concentration and a possible dilution factor prior to using a quantitative method such as the Spectroquant® test.
All our Cell and Reagent Test Kits are equipped with the unique Live ID (2D barcode) which allows seamless method recognition and contains essential information such as lot number, expiry date, and automatic calibration updates.
The Spectroquant® Reagent Test Kits contain highly stable, ready-to-use reagent mixtures to perform the analysis according to the procedure described in the accompanying instruction leaflet.

Legal Information

MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1 - STOT RE 2 Oral - STOT SE 3

Target Organs

Respiratory system, Thyroid

Storage Class Code

3 - Flammable liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Photometric determination with Dimethylglyoxime subsequent to acid mineralisation; DIN decomposition

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service