Skip to Content
Merck

Skip To

1.13136

Hydrochloric acid solution

c(HCl) = 0.357 mol/l (1/2.8 N), ready-to-use volumetric solution for titration, Titripur®

Synonym(s):

Hydrochloric acid solution, Hydrogen chloride solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
10 L
Check Cart for Availability
€ 186,00

About This Item

Empirical Formula (Hill Notation):
HCl
CAS Number:
Molecular Weight:
36.46
NACRES:
NA.21
UNSPSC Code:
12352301
MDL number:

€ 186,00


Check Cart for Availability
This product features our Titripac® packaging technology to reduce the risk of contamination.

Request more information
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Hydrochloric acid solution, c(HCl) = 0.357 mol/l (1/2.8 N), Titripur®

Quality Segment

product line

Titripur®

form

liquid

quality

Analyzed in our ISO 17025 accredited QC lab

reaction suitability

reaction type: Acid-base reactions

packaging

Titripac® of

concentration

0.3571 M

technique(s)

titration: suitable

pH

1.2 (20 °C in H2O)

density

1.01 g/cm3 at 20 °C

storage temp.

15-25°C

SMILES string

Cl

InChI

1S/ClH/h1H

InChI key

VEXZGXHMUGYJMC-UHFFFAOYSA-N

Features and Benefits

Accreditation:
This volumetric solution is analyzed by our calibration laboratory D-K-15185-01-00 which is accredited according to DIN EN ISO/IEC 17025 for analysis of amount-of-substance concentrations in volumetric solutions by DAkkS (Deutsche Akkreditierungsstelle - German National Accreditation Body). The accreditation certificate can be found at www.sigmaaldrich.com/ISO17025.

Packaging

SKU-pack size numbers ending with a "3" are SmartChemicals with an RFID tag on the label for seamless data transfer to the instrument, e.g. xxxxxx1003 or xxxxxxx 4003. More information on www.sigmaaldrich.com/SmartChemicals

Titripac® packaging - all SKU-pack size numbers ending "4000, 4003, 9010, 9013" - more information on www.sigmaaldrich.com/Titripac

Analysis Note

Form liquid
Amount-of-substance concentration 0.3553 - 0.3589 mol/L
Measurement uncertainty ± 0.0011 mol/L
Traceability NIST SRM

The concentration is determined by volumetric titration and refers to 20°C.
The amount-of-substance concentration of this volumetric solution is traceable to a primary standard reference material (SRM) from the National Institute of Standards and Technology, Gaithersburg, USA (NIST SRM 723 Tris(hydroxymethyl)aminomethane) by means of volumetric standard Tris(hydroxymethyl)aminomethane (article number 1.02408), certified reference material according to ISO 17034, analyzed by our accredited calibration laboratory of Merck KGaA, Darmstadt, Germany according to DIN EN ISO/IEC 17025. The uncertainty is expressed as expanded measurement uncertainty with a coverage factor k=2 covering a confidence level of 95%.

Note: The titer is a correction factor to correct for variations of the volumetric solution, the titration equipment, the temperature and other laboratory conditions. For correct titration results it is recommended to determine a titer with the laboratory specific equipment and under laboratory specific conditions directly after opening a new bottle and at regular time intervals.

Legal Information

Titripac is a registered trademark of Merck KGaA, Darmstadt, Germany
Titripur is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1.090581.090601.09057
reaction suitability

reaction type: Acid-base reactions

reaction suitability

reaction type: Acid-base reactions

reaction suitability

reaction type: Acid-base reactions

reaction suitability

reaction type: Acid-base reactions

technique(s)

titration: suitable

technique(s)

titration: suitable

technique(s)

titration: suitable

technique(s)

titration: suitable

product line

Titripur®

product line

Titripur®

product line

Titripur®

product line

Titripur®

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

pH

1.2 (20 °C in H2O)

pH

<1 (20 °C in H2O)

pH

1.2 (20 °C in H2O)

pH

<1 (20 °C in H2O)

packaging

Titripac® of

packaging

Titripac® of

packaging

Titripac® of

packaging

Titripac® of


Still not finding the right product?

Explore all of our products under Hydrochloric acid solution


pictograms

Corrosion

signalword

Warning

hcodes

Hazard Classifications

Met. Corr. 1

flash_point_f

Not applicable

flash_point_c

Not applicable

Storage Class

12 - Non Combustible Liquids



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Articles

Discover the ChemiSphere™ App for secure, contactless reagent data transfer to Metrohm OMNIS titrators, ensuring accuracy, traceability, and efficiency.

Related Content

SmartChemicals enable wireless CoA data transfer, reducing human error in titration experiments.





Global Trade Item Number

SKUGTIN
113136901304061841064800
113136901004022536147314

Questions

Reviews

No rating value

Active Filters