Skip to Content
Merck

Skip To

1.11471

Nutrient agar

NutriSelect® Prime, acc. to German Standard Methods, granular, for heterotrophic bacteria, pkg of 500 g, pkg of 5 kg

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
500 g
Check Cart for Availability
€ 372,00
5 kg
Check Cart for Availability
€ 3.560,00

About This Item

NACRES:
NA.74
UNSPSC Code:
41106213

€ 372,00


Check Cart for Availability

Request more information
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


agency

acc. to German Standard Methods

Quality Segment

form

granular

packaging

pkg of 5 kg, pkg of 500 g

manufacturer/tradename

NutriSelect® Prime

technique(s)

microbiological culture: suitable

pH

7.1-7.5 (37 °C, 40 g/L in H2O, after autoclaving)

solubility

40 g/L

bulk density

560 kg/m3

application(s)

food and beverages
microbiology

storage temp.

15-25°C

suitability

heterotrophic bacteria

Analysis Note

Appearance (clearness): clear
Appearance (color): yellowish brown
pH-value (25 °C): 7.1 - 7.5
Growth promotion test in accordance with the current version of DIN EN ISO 11133.

Inoculum on reference medium (Bacillus subtilis ATCC 6633 (WDCM 00003)):
Inoculum on reference medium (Escherichia coli ATCC 8739 (WDCM 00012)):
Inoculum on reference medium (Escherichia coli ATCC 25922 (WDCM 00013)):
Colony count (Bacillus subtilis ATCC 6633 (WDCM 00003)):
Colony count (Escherichia coli ATCC 8739 (WDCM 00012)):
Colony count (Escherichia coli ATCC 25922 (WDCM 00013)):
Recovery on test medium (Bacillus subtilis ATCC 6633 (WDCM 00003)): ≥ 70 %
Recovery on test medium (Escherichia coli ATCC 8739 (WDCM 00012)): ≥ 70 %
Recovery on test medium (Escherichia coli ATCC 25922 (WDCM 00013)): ≥ 70 %
Incubation: 44 ± 4 hours at 36 ± 1 °C, aerobic
A recovery rate of 70 % is equivalent to a productivity value of 0.7.
The indicated colony counts result from the sum of a triple determination.
Reference media: Tryptic Soy Agar

Other Notes

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1.108661.0386404163
suitability

heterotrophic bacteria

suitability

selective for yeasts

suitability

nonselective for

suitability

nonselective for bacteria (General Media)

technique(s)

microbiological culture: suitable

technique(s)

microbiological culture: suitable

technique(s)

microbiological culture: suitable

technique(s)

microbiological culture: suitable

agency

acc. to German Standard Methods

agency

according to ISO 10718

agency

-

agency

according to ISO 6579-1:2017

form

granular

form

granular

form

powder

form

powder

application(s)

food and beverages
microbiology

application(s)

food and beverages
microbiology

application(s)

food and beverages
pharmaceutical

application(s)

agriculture
bioburden testing
environmental
food and beverages
life science and biopharma
water monitoring, microbiology

pH

7.1-7.5 (37 °C, 40 g/L in H2O, after autoclaving)

pH

5.3-5.7 (25 °C, 77 g/L in H2O, after autoclaving)

pH

7.5±0.2 (25 °C)

pH

-


Still not finding the right product?

Explore all of our products under Nutrient agar


Storage Class

11 - Combustible Solids

wgk

WGK 3



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library





Global Trade Item Number

SKUGTIN
111471500004022536133034
111471050004022536133010

Questions

Reviews

No rating value

Active Filters