Skip to Content
Merck
All Photos(1)

Key Documents

1.10047

Supelco

Total hardness Test Strips, colorimetric

individually sealed, water hardness 5-25 °d, water hardness 6-31 °e, water hardness 9-45 °f, MQuant®

Synonym(s):

Water Hardness Chemical Test Strips

Sign Into View Organizational & Contract Pricing

Select a Size

25000 STRIPS
€5,790.00

€5,790.00


Estimated to ship on23 May 2025


Request a Bulk OrderRequest more information

Select a Size

Change View
25000 STRIPS
€5,790.00

About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

€5,790.00


Estimated to ship on23 May 2025


Request a Bulk OrderRequest more information

Product Name

Total hardness Test, colorimetric, water hardness 5-25 °d, water hardness 6-31 °e, water hardness 9-45 °f, MQuant®

product line

MQuant®

Quality Level

specific analyte(s)

calcium, magnesium

water hardness measuring range

5-25 °d
6-31 °e
9-45 °f

detection method

colorimetric

storage temp.

15-25°C

General description

Method: colorimetric with test strips (individually sealed) >5 - >10 - >15 - >20 - >25°d >6 - >13 - >19 - >25 - >31°e >9 - >18 - >27 - >36 - >45°f MQuant®

Legal Information

MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service