1.09526
pH-indicator paper pH 1 - 10 Universal indicator
roll L 4.8 m, with color scale, graduations and accuracy: 1 pH unit, pH range 1-10, MQuant®
Synonym(s):
pH chemical test paper paper pH 1 - 10 Universal indicator, pH test paper paper pH 1 - 10 Universal indicator
About This Item
Recommended Products
product name
pH-indicator paper pH 1 - 10 Universal indicator, roll L 4.8 m , with color scale, graduations and accuracy accuracy: 1 pH unit, pH range 1-10, MQuant®
product line
MQuant®
Quality Level
feature
non-bleeding: no
graduations and accuracy measuring range
accuracy: 1 pH unit
roll L
4.8 m , with color scale
pH range
1-10
detection method
colorimetric
storage temp.
15-25°C
Related Categories
General description
Analysis Note
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service