Skip to Content
Merck
All Photos(2)

Key Documents

1.09514

Supelco

pH-indicator paper Congo red paper

roll L 4.8 m , with color scale, reag. Ph. Eur., MQuant®

Synonym(s):

pH chemical test paper paper Congo red paper, pH test paper paper Congo red paper

Sign Into View Organizational & Contract Pricing

Select a Size

3 ROLLS
€52.20

€52.20


Estimated to ship on21 May 2025


Request a Bulk OrderRequest more information

Select a Size

Change View
3 ROLLS
€52.20

About This Item

UNSPSC Code:
41113034
NACRES:
NB.35

€52.20


Estimated to ship on21 May 2025


Request a Bulk OrderRequest more information

Agency

reag. Ph. Eur.

Quality Level

product line

MQuant®

feature

non-bleeding: no

roll L

4.8 m , with color scale

detection method

colorimetric

storage temp.

15-25°C

General description

MQuant® Congo red paper is suitable to monitor a qualitative pH change in the acidic range. The paper changes its color from blue-violet (pH < 2) to red-orange (pH > 5). pH indicator papers supplied in rolls are better suited for longterm storage, since they are protected against external influences. pH indicator papers are manufactured by impregnating high-quality filter paper with indicator or mixed-indicator solutions. Therefore, the paper is dryed, cut and packed into the corresponding sizes. For a semi-quantitative monitoring of the pH value, please refer to our MQuant® pH-indicator papers or strips and for a quantitative measurement to our Reflectoquant® test strips.

Analysis Note

Application test: conforms

Legal Information

MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
REFLECTOQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service