Skip to Content
Merck
All Photos(1)

Key Documents

1.09362

Millipore

LiChroprep® RP-8 (40-63 µm)

for liquid chromatography

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
€832.00
1 KG
€3,004.00

€832.00


Estimated to ship on26 May 2025



Select a Size

Change View
250 G
€832.00
1 KG
€3,004.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
41115711

€832.00


Estimated to ship on26 May 2025


material

silica gel (irregularly-shaped)

Quality Level

product line

LiChroPrep®

matrix active group

Irregular-Shaped Silica Gel

particle size

40-63 μm

pore size

10 nm mean pore size

bulk density

570 kg/m3

application(s)

plasma purification

separation technique

reversed phase

storage temp.

no temp limit

General description

Irregular-shaped Silica gel with narrow, well defined particle size distribution delivering high separating performance, permeability and good reproducibility

Features and Benefits

LiChroprep® RP-8 (40-63 µm) provides users with:

  • Reproducible, homogeneous, irregularly shaped silica gel matrix
  • Narrow, well defined particle size distribution for high performance and high permeability
  • Large batch sizes up to 200 kg in the modification

Packaging

Plastic bottle

Analysis Note

Application test: passes test
Pressure: ≤ 10 bar
Theoretical Plates (N/m) (Toluene): ≥ 1500
particle size (Malvern) (d10): 40 - 57 µm
particle size (Malvern) (d50): 60 - 75 µm
particle size (Malvern) (d90): 90 - 115 µm

Legal Information

LICHROPREP is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service