Skip to Content
Merck
All Photos(1)

Key Documents

60756

Supelco

C₁₈-Reversed Phase Silica Gel Chromatography Media

550-950 mesh, fully endcapped

Synonym(s):

C18-reversed phase silica gel, Reversed phase silica gel C18

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
€405.00
250 G
€1,880.00

€405.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
50 G
€405.00
250 G
€1,880.00

About This Item

EC Number:
MDL number:
UNSPSC Code:
12000000
NACRES:
SB.52

€405.00


Estimated to ship on28 May 2025


Request a Bulk Order

Product Name

C18-Reversed phase silica gel, for column chromatography, fully end-capped

grade

for column chromatography

form

gel form

quality

fully end-capped

technique(s)

LPLC: suitable

surface area

380 m2/g

matrix

Silica

matrix active group

C18 phase

particle size

15-25 μm

pore size

100 Å pore size

suitability

suitable for column chromatography

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

Application

for reversed phase chromatography; designed for special tuning of selectivity

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service