Skip to Content
Merck
All Photos(1)

Key Documents

60759

Supelco

Silica gel C₈-Reversed phase Inorganic Sorbent

230-400 mesh, fully endcapped

Synonym(s):

Reversed phase silica gel C8

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
€337.00

€337.00


Estimated to ship on25 May 2025


Request a Bulk Order

Select a Size

Change View
50 G
€337.00

About This Item

EC Number:
MDL number:
UNSPSC Code:
12000000
NACRES:
SB.52

€337.00


Estimated to ship on25 May 2025


Request a Bulk Order

Product Name

Silica gel C8-Reversed phase, for column chromatography, fully endcapped

grade

for column chromatography

Quality Level

form

powder

quality

fully endcapped

technique(s)

LPLC: suitable

matrix

Silica

matrix active group

C8

particle size

40-63 μm

pore size

90 Å pore size

suitability

suitable for column chromatography

separation technique

reversed phase

General description

Reversed-phase silica gels are generally suitable to retain catalysts by hydrophobic interactions.[1]

Application

Silica gel C8-reversed phase was used in the purification of tertiary sulphonamide using FSPE.[2] Pressure induced C8 reversed phase silica gel was synthesized through a sol-gel templating technique.[3]

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effective viscosity of glycerin in a nanoporous silica gel.
Han, Aijie, et al.
Journal of Applied Physics, 104, 124904-124904 (2008)
Fluorous scavenger for parallel preparation of tertiary sulfonamides leading to secondary amines.
Basle, Emmanuel, et al.
Tetrahedron Letters, 48, 8138-8140 (2007)
J Horn et al.
Topics in current chemistry, 242, 43-75 (2004-01-01)
Catalysts immobilized on solid supports have become valuable tools for simplified product isolation and catalyst recycling. An alternative to covalent attachment to a solid support is to immobilize catalysts by non-covalent bonding through hydrogen bridges, or ionic, hydrophobic or fluorous

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service