Skip to Content
Merck
All Photos(1)

Documents

1.09127

Millipore

Tripod Adapter

for use with MAS-100 Eco®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116500
NACRES:
JA.14

material

aluminum adapter

description

connector

product line

MAS-100®

feature

autoclavable

packaging

pkg of 1 unit

manufacturer/tradename

MAS-100 Eco®

application(s)

air monitoring
food and beverages

compatibility

for use with MAS-100 Eco®

storage temp.

room temp

General description

Tripod adapter is set to be mounted to MAS-100 Eco® air sampler and tripod to allow quick attachment of the air sampler to the tripod without repeated screwing. The air sampler must be stably attached to the tripod, therefore a useful addition for the operation of the MAS-100 Eco® on the tripod is the tripod adapter as it ensures a stable interaction. The tripod adapter is the ideal accessory for the MAS-100 Eco® microbial air sampler. It is characterized by an easy operation and can be used without tools. The user must regularly check that the connection from the tripod to the adapter is stable and ensure to tighten the screw of the adapter.

Application

Microbial air sampling devices are extensively used in Pharmaceutical and Personal care industries as well as in Food, Beverage and Environmental applications.

Linkage

Replaces: 1.09127.0002

Legal Information

MAS-100 is a registered trademark of MBV AG
MAS-100 Eco is a registered trademark of MBV AG

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service