Key Documents
1.17269
Millipore
Battery Charger
for use with MAS-100 Eco®
Select a Size
1172690001
€194.00
€194.00
Estimated to ship on28 May 2025
Request more information
Select a Size
Change View
1172690001
€194.00
About This Item
UNSPSC Code:
41103403
NACRES:
NA.74
€194.00
Estimated to ship on28 May 2025
Request more information
material
plastic hardware
Agency
non-compendial
product line
MAS-100®
feature
electronic
manufacturer/tradename
MAS-100 Eco®
application(s)
air monitoring
cosmetics
food and beverages
pharmaceutical
compatibility
for use with MAS-100 Eco®
General description
The MAS-100 Eco® battery charger is an additional accessory for MAS-100 Eco® microbial air sampler. The battery charger for MAS-100 Eco® air sampler has an input voltage of 100 to 240 V/AC (47-63 Hz) and an output voltage of 5 V/DC (500 mA)
Application
The MAS-100 Eco® charger is an additional accessory for MAS-100 Eco® air sampler that is used for microbial monitoring of ambient air in controlled environments. Microbial air sampling devices are extensively used in Food, Beverage and Environmental applications.
Legal Information
MAS-100 is a registered trademark of MBV AG
MAS-100 Eco is a registered trademark of MBV AG
accessory