Skip to Content
Merck
All Photos(1)

Key Documents

1.09223

Millipore

Tripod quick change adapter

MAS-100® Family

Sign Into View Organizational & Contract Pricing

Select a Size

1 UNIT
€627.00

€627.00


Estimated to ship on01 June 2025


Request more information

Select a Size

Change View
1 UNIT
€627.00

About This Item

UNSPSC Code:
41103403
NACRES:
NA.74

€627.00


Estimated to ship on01 June 2025


Request more information

material

aluminum adapter

Agency

non-compendial

description

connector

feature

adjustable

packaging

pkg of 1 kit

manufacturer/tradename

MAS-100® Family

application(s)

air monitoring
cosmetics
food and beverages
pharmaceutical

compatibility

for use with MAS-100 NT®
for use with MAS-100 VF®

storage temp.

no temp limit

General description

Tripod quick change adapter for all portable MAS-100® air sampler, with exception of MAS-100 Eco® set to be mounted to MAS-100® air sampler and tripod to allow quick attachment of the air sampler to the tripod without repeated screwing. The air sampler must be stably attached to the tripod, therefore a useful addition for the operation of the MAS-100® on the tripod is the quick change adapter as it not only ensures a stable interaction but also eliminates the need to repeatedly screw the unit to the tripod and helps to reduce the wear and tear of the screws on the bottom of the unit. The quick-change adapter is the ideal accessory for the MAS-100® microbial air sampler. It is characterized by an easy operation and can be used without tools. The adapter plate made of aluminum with a rubber mat is designed to remain permanently attached to the air sampler and the quick-change adapter made of solid hard plastic is screwed on the tripod socket. A simple click of the quick-change adapter plate into the adapter enables the air sampler to mount tightly to the mobile and height adjustable tripod. The user must regularly check that the connection from the tripod to the plate is stable and ensure to tighten the screw of the adapter plate.

Application

Microbial air sampling devices are extensively used in Pharmaceutical and Personal care industries as well as in Food, Beverage and Environmental applications.

Legal Information

MAS-100 is a registered trademark of MBV AG
MAS-100 Eco is a registered trademark of MBV AG
MAS-100 NT is a registered trademark of MBV AG
MAS-100 VF is a registered trademark of MBV AG

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service