Skip to Content
Merck
All Photos(1)

Key Documents

1.09086

Supelco

Barium perchlorate solution

c(Ba(ClO₄)₂) = 0.005 mol/l in 2-propanol/water (80/20), ready-to-use volumetric solution for titration, Titriplex®

Synonym(s):

Barium perchlorate solution in 2-propanol/water; Bariumperchlorate solution in Isoproanol/water

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
€93.80
1 L
€99.80

€93.80


Estimated to ship on25 May 2025


Request more information

Select a Size

Change View
1 L
€93.80
1 L
€99.80

About This Item

UNSPSC Code:
12352302
NACRES:
NA.21

€93.80


Estimated to ship on25 May 2025


Request more information

Product Name

Barium perchlorate solution, in 2-propanol/water (80/20) c(Ba(ClO4)2) = 0.005 mol/l, Titripur®

Quality Level

product line

Titripur®

form

liquid

quality

Analyzed in our ISO 17025 accredited QC lab

reaction suitability

reaction type: Redox Reactions

concentration

0.005 M

technique(s)

titration: suitable

pH

3.3 (20 °C in H2O)

transition temp

flash point 21 °C

density

0.85 g/cm3 at 20 °C

storage temp.

15-25°C

Application


  • L-serine restored lysosomal failure in cells derived from patients with BPAN reducing iron accumulation with eliminating lipofuscin.: This study involves the use of a catalytic system based on 2,2′-bipyridine for biochemical applications related to neurodegenerative diseases, focusing on cellular repair and iron homeostasis (Mun et al., 2024).


Features and Benefits

Accreditation:
This volumetric solution is analyzed by our calibration laboratory D-K-15185-01-00 which is accredited according to DIN EN ISO/IEC 17025 for analysis of amount-of-substance concentrations in volumetric solutions by DAkkS (Deutsche Akkreditierungsstelle - German National Accreditation Body). The accreditation certificate can be found at www.sigmaaldrich.com/ISO17025.

Packaging

SKU-pack size numbers ending with a "3" are SmartChemicals with an RFID tag on the label for seamless data transfer to the instrument, e.g. xxxxxx1003 or xxxxxxx 4003. More information on www.sigmaaldrich.com/SmartChemicals

Analysis Note

Form liquid
Amount-of-substance concentration 0.004950 - 0.005050 mol/L
Measurement uncertainty ± 0.000040 mol/L
Traceability NIST SRM

The concentration is determined by volumetric titration and refers to 20°C.
The amount-of-substance concentration of this volumetric solution is determined by back-titration with copper(II) nitrate solution (article number 1.02753), standardized by Titriplex(III) solution (article number 1.08431) which has been standardized against volumetric standard zinc (article number 1.02409).

This volumetric solution is traceable to a primary standard reference material (SRM) from the National Institute of Standards and Technology, Gaithersburg, USA (NIST SRM 682 zinc) by means of volumetric standard zinc (article number 1.02409), certified reference material according to ISO 17034, analyzed by our accredited calibration laboratory of Merck KGaA, Darmstadt, Germany according to DIN EN ISO/IEC 17025. The uncertainty is expressed as expanded measurement uncertainty with a coverage factor k=2 covering a confidence level of 95%.

Note: The titer is a correction factor to correct for variations of the volumetric solution, the titration equipment, the temperature and other laboratory conditions. For correct titration results it is recommended to determine a titer with the laboratory specific equipment and under laboratory specific conditions directly after opening a new bottle and at regular time intervals.

Legal Information

Titripur is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

69.8 °F

Flash Point(C)

21 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service