Skip to Content
Merck
All Photos(2)

Key Documents

1.04818

Supelco

Potassium chloride solution

c(KCl) = 3 mol/l, saturated with silver chloride, for filling of electordes

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€38.10

€38.10


Check Cart for Availability


Select a Size

Change View
100 ML
€38.10

About This Item

Linear Formula:
KCl
CAS Number:
Molecular Weight:
74.55
Beilstein:
1711999
MDL number:
UNSPSC Code:
12352402
NACRES:
NA.21
grade:
for analytical purposes
form:
liquid

€38.10


Check Cart for Availability

Product Name

Potassium chloride solution, saturated with silver chloride 3 mol/l

grade

for analytical purposes

Quality Level

form

liquid

reaction suitability

reaction type: Complexometric reactions

technique(s)

titration: suitable

pH

6.2 (20 °C in H2O)

bp

100 °C/1013 hPa

density

1.13 g/cm3 at 20 °C

storage temp.

15-25°C

SMILES string

[Cl-].[K+]

InChI

1S/ClH.K/h1H;/q;+1/p-1

InChI key

WCUXLLCKKVVCTQ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application


  • Amperometric lactate biosensor for flow injection analysis based on a screen-printed carbon electrode containing Meldola′s Blue-Reinecke salt, coated with lactate dehydrogenase and NAD+.: This study explores the development of an amperometric lactate biosensor using a potassium chloride solution to enhance the performance of the electrode, providing a novel application in analytical chemistry for precise lactate measurement (Piano et al., 2010).

  • Stability improvement of alpha-amylase entrapped in kappa-carrageenan beads: physicochemical characterization and optimization using composite index.: This research investigates the stability of alpha-amylase in kappa-carrageenan beads using potassium chloride solution, demonstrating its effectiveness in maintaining enzyme activity in analytical procedures (Sankalia et al., 2006).

  • Volume osmotic flows of non-homogeneous electrolyte solutions through horizontally mounted membrane.: This paper discusses the role of potassium chloride solution in studying volume osmotic flows, highlighting its application in the analysis of electrolyte behavior in analytical chemistry (Slezak et al., 2002).

  • Microglial reaction in the rat cerebral cortex induced by cortical spreading depression.: The study uses potassium chloride solution to induce cortical spreading depression in rats, providing insights into the microglial reaction and contributing to the understanding of cerebral cortex processes in analytical chemistry contexts (Gehrmann et al., 1993).

  • Possible factors in the adhesion of fissure sealants to enamel.: This research investigates the factors affecting the adhesion of fissure sealants to enamel, with potassium chloride solution playing a critical role in the experimental setup, offering valuable data for dental material analysis (Williams & von Fraunhofer, 1979).

Analysis Note

concentration (KCl): 2.97 - 3.03 mol/l
Ag (Silver): ≥ 0.16 g/l

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service