Skip to Content
Merck
All Photos(1)

Key Documents

1.08445

Supelco

Ethylenedinitrilotetraacetic acid tripotassium salt dihydrate

suitable for titration

Synonym(s):

Ethylenedinitrilotetraacetic acid tripotassium salt dihydrate

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
€259.00

€259.00


Estimated to ship on19 May 2025


Request more information

Select a Size

Change View
250 G
€259.00

About This Item

Empirical Formula (Hill Notation):
C10H13K3N2O8 · 2H2O
CAS Number:
Molecular Weight:
442.54
MDL number:
UNSPSC Code:
12352107
EC Index Number:
241-543-5
NACRES:
NA.21

€259.00


Estimated to ship on19 May 2025


Request more information

Product Name

Ethylenedinitrilotetraacetic acid tripotassium salt dihydrate, Ethylenedinitrilotetraacetic acid tripotassium salt dihydrate. CAS 65501-24-8, pH 8 - 9 (50 g/l, H₂O, 20 °C).

Quality Level

form

solid

reaction suitability

reaction type: Complexometric reactions

technique(s)

titration: suitable

pH

8-9 (20 °C, 50 g/L in H2O)

mp

180-185 °C

storage temp.

no temp limit

SMILES string

[K+].[K+].[K+].[N+H](CCN(CC(=O)[O-])CC(=O)[O-])(CC(=O)[O-])CC(=O)[O-].O.O

InChI

1S/C10H16N2O8.3K.2H2O/c13-7(14)3-11(4-8(15)16)1-2-12(5-9(17)18)6-10(19)20;;;;;/h1-6H2,(H,13,14)(H,15,16)(H,17,18)(H,19,20);;;;2*1H2/q;3*+1;;/p-3

InChI key

MAPFUJCWRWFQIY-UHFFFAOYSA-K

Analysis Note

Assay (complexometric, calc. on anhydrous substance): ≥ 99 %
Identity (IR-spectrum): passes test
Identity (Potassium): passes test
pH (50 g/l; water): 7.0 - 9.0
Chloride (Cl): ≤ 0.01 %
Heavy metals (as Pb): ≤ 0.002 %
Fe (Iron): ≤ 0.001 %
Water (according to Karl Fischer): 7 - 9 %

Pictograms

Exclamation markHealth hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - STOT RE 2 Inhalation

Target Organs

Respiratory Tract

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service