Skip to Content
Merck
All Photos(1)

Key Documents

60153

Sigma-Aldrich

Potassium citrate tribasic monohydrate

purum p.a., ≥99.0% (NT)

Synonym(s):

Citric acid tripotassium salt, Tripotassium citrate

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
€78.80
1 KG
€128.00

€78.80


Estimated to ship on30 March 2025


Request a Bulk Order

Select a Size

Change View
500 G
€78.80
1 KG
€128.00

About This Item

Linear Formula:
HOC(COOK)(CH2COOK)2 · H2O
CAS Number:
Molecular Weight:
324.41
Beilstein:
3924344
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21
Assay:
≥99.0% (NT)
form:
liquid
solubility:
H2O: 0.5 g/10 mL, clear, colorless

€78.80


Estimated to ship on30 March 2025


Request a Bulk Order

grade

purum p.a.

Quality Level

Assay

≥99.0% (NT)

form

liquid

pH

7.5-9.0 (25 °C, 50 mg/mL in H2O)

mp

275 °C (dec.) (lit.)

solubility

H2O: 0.5 g/10 mL, clear, colorless

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Ca: ≤50 mg/kg
Cd: ≤50 mg/kg
Co: ≤50 mg/kg
Cu: ≤50 mg/kg
Fe: ≤50 mg/kg
Na: ≤1500 mg/kg
Ni: ≤50 mg/kg
Pb: ≤50 mg/kg
Zn: ≤50 mg/kg

SMILES string

O.[K+].[K+].[K+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O

InChI

1S/C6H8O7.3K.H2O/c7-3(8)1-6(13,5(11)12)2-4(9)10;;;;/h13H,1-2H2,(H,7,8)(H,9,10)(H,11,12);;;;1H2/q;3*+1;/p-3

InChI key

PJAHUDTUZRZBKM-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Potassium citrate tribasic monohydrate is the tripotassium salt of citric acid. It has been proposed as a promising organic salt for the formation of aqueous two-phase system (ATPS). UCON (temperature-responsive polymer) + potassium citrate ATPS has been studied.[1]

Application

Potassium citrate tribasic monohydrate (tribasic potassium citrate) may be added as a promoter during the preparation of activated carbon-supported CuCl2 + PdCl2 + NaOH catalyst. This catalyst was employed for the conversion of ethanol to diethyl carbonate (DEC).1 It may be used to compose the fermentation medium for Aspergillus niger 72-4.[2] It may be used as a standard to investigate the influence of Bergenia ligulata rhizome (BLR) on CaC2O4 crystallization.[3]

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carbon dioxide fixation by cell-free extracts of Aspergillus niger.
Woronick CL and Johnson MJ.
The Journal of Biological Chemistry, 235(1), 9-15 (1960)
(Liquid+ liquid) equilibria of polymer-salt aqueous two-phase systems for laccase partitioning: UCON 50-HB-5100 with potassium citrate and (sodium or potassium) formate at 23?C.
Lladosa E, et al.
The Journal of Chemical Thermodynamics, 55, 166-171 (2012)
Antiurolithic effect of Bergenia ligulata rhizome: an explanation of the underlying mechanisms.
Bashir S and Gilani AH.
Journal of Ethnopharmacology, 122(1), 106-116 (2009)
Clay Mechlin et al.
Journal of endourology, 25(9), 1541-1545 (2011-08-06)
Oral citrate supplements have been shown to decrease kidney stone recurrence rates in both laboratory and clinical studies. The taste of the citrate supplements, however, is poor, and long-term compliance is low. Our objective was to determine if Splenda(®) added
Donna J Claes et al.
Pediatric nephrology (Berlin, Germany), 27(11), 2031-2038 (2012-01-28)
Cystinuria is a relatively uncommon cause of pediatric stone disease, but has significant morbidity if not properly controlled because of its significant stone recurrence rate. Cystinuria is caused by the inability of the renal tubules to reabsorb filtered cystine, which

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service