Skip to Content
Merck
All Photos(1)

Documents

03660

Sigma-Aldrich

Ethylenediaminetetraacetic acid dipotassium salt dihydrate

puriss. p.a., ≥99.0% (KT)

Synonym(s):

K2EDTA, (Ethylenedinitrilo)tetraacetic acid dipotassium salt, Dipotassium ethylenediaminetetraacetate dihydrate, EDTA dipotassium salt, Edathamil, Potassium ethylenediaminetetraacetate dibasic

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOOCCH2(KOOCCH2)NCH2CH2N(CH2COOK)CH2COOH · 2H2O
CAS Number:
Molecular Weight:
404.45
EC Number:
UNSPSC Code:
12352100
NACRES:
NA.21

grade

puriss. p.a.

Quality Level

Assay

≥99.0% (KT)

pH

4-5 (25 °C, 50 mg/mL in H2O)

mp

255-280 °C (dec.) (lit.)

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤200 mg/kg

cation traces

Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Na: ≤1000 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

SMILES string

O.O.O.O.[K+].[K+].[K+].[K+].OC(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC(O)=O.OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O

InChI

1S/C10H16N2O8.2K.2H2O/c13-7(14)3-11(4-8(15)16)1-2-12(5-9(17)18)6-10(19)20;;;;/h1-6H2,(H,13,14)(H,15,16)(H,17,18)(H,19,20);;;2*1H2/q;2*+1;;/p-2

InChI key

JFROQFOCFOKDKU-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Ethylenediaminetetraacetic acid dipotassium salt dihydrate (K2EDTA), is a well known chelating agent with various uses in chemical synthesis, analytical chemistry, and biochemistry. In chemical synthesis, K2EDTA is often employed to remove metal impurities from solutions or as a complexation agent to control the reactivity of metal ions. It can be used to sequester or chelate metal ions such as calcium, magnesium, iron, and copper, which helps prevent undesired side reactions.

Application

Ethylenediaminetetraacetic acid dipotassium salt dihydrate (K2EDTA) can be used as:
  • A mobile phase modifier in the separation of UV filters using HPLC.
  • A chelating agent in the determination of traces of beryllium compound by colorimetric method.

Used to eliminate inhibition of enzyme catalyzed reactions due to traces of heavy metals

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Inhalation

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Onsite direct-read system for semi-quantitative detection of traces of beryllium on surfaces
Tekleab TM, et al.
Journal of Environmental Monitoring, 8(6), 625-629 (2006)
Mark A Yondola et al.
Journal of virology, 81(8), 4264-4271 (2007-02-09)
One of the most interesting functions attributed to the adenovirus early region 4 open reading frame 3 (E4 ORF3) protein is its reorganization of promyelocytic leukemia (PML) protein nuclear bodies. These normally punctate structures are reorganized by E4 ORF3 into
Determination of sixteen UV filters in suncare formulations by high-performance liquid chromatography.
Schakel DJ, et al.
Journal of Chromatography A, 1049(1-2), 127-130 (2004)
Lisa K Philp et al.
PloS one, 10(2), e0117494-e0117494 (2015-02-07)
High saturated fat (HF-S) diets increase intramyocellular lipid, an effect ameliorated by omega-3 fatty acids in vitro and in vivo, though little is known about sex- and muscle fiber type-specific effects. We compared effects of standard chow, HF-S, and 7.5%

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service