Skip to Content
Merck
All Photos(1)

Key Documents

1.07754

Millipore

Silica gel 60

extra pure for column chromatography 0.063 - 0.200 mm (70 - 230 mesh ASTM)

Synonym(s):

Silica, Silicon dioxide

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
€248.00
1 KG
€441.00

€248.00


Estimated to ship on23 May 2025



Select a Size

Change View
500 G
€248.00
1 KG
€441.00

About This Item

Linear Formula:
SiO2
CAS Number:
Molecular Weight:
60.08
MDL number:
UNSPSC Code:
41115711
EC Index Number:
231-545-4

€248.00


Estimated to ship on23 May 2025


material

silica gel (irregularly-shaped)

Quality Level

matrix active group

Irregular-Shaped Silica Gel

particle size

63-200 μm

pore size

6 nm mean pore size

pH

7 (20 °C, 100 g/L in H2O, slurry)

bp

2230 °C (lit.)

mp

>1600 °C (lit.)

bulk density

200‑1430 kg/m3

application(s)

plasma purification
plasma purification

storage temp.

no temp limit

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

irregularly shaped silica gel matrix ideal for simple adsorption processes and normal-phase chromatography

Features and Benefits

Silica gel 60 provides users with:

  • High adsorptive capacity
  • High chemical inertness and non-toxicity.
  • Protective moisture-sensitive substances during storage and transport or for maintaining the dryness of anhydrous solvents.
  • Granulates in different sizes or beads

Packaging

Plastic bottle

Analysis Note

pH-value (10 % suspension): 6.0 - 7.5
Chloride (Cl): ≤ 0.008 %
Sulfate (SO₄): ≤ 0.003 %
Cd (Cadmium): ≤ 0.0005 %
Cu (Copper): ≤ 0.0005 %
Fe (Iron): ≤ 0.002 %
Pb (Lead): ≤ 0.0005 %
Zn (Zinc): ≤ 0.0005 %
Particle size: 90% between 0.063-0.200 mm
Loss on drying (150 °C): ≤ 7.0 %
UV absorption (Dichloro methane extract): conforms

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service