Skip to Content
Merck
All Photos(2)

Documents

1.07730

Supelco

Silica gel 60 GF254

for thin-layer chromatography

Synonym(s):

Silica gel

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41115711
EC Index Number:
231-545-4
NACRES:
NB.21

material

silica gel 60 matrix

Quality Level

form

powder

potency

>5000 mg/kg LD50, skin (Rabbit)

feature

binder Gypsum
fluorescent indicator

packaging

pkg of 25 kg

technique(s)

thin layer chromatography (TLC): suitable

particle size

5-40 μm

pore size

60 Å medium pore diameter

pH

7 (20 °C, 100 g/L in H2O, slurry)

bp

2230 °C

mp

>1600 °C

bulk density

200‑800 kg/m3

storage temp.

no temp limit

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Standardized sorbents ensure reliable results every time. Our broad spectrum meets all kinds of needs. In TLC, silica gel 60 sorbent is considered the most versatile. Our assortment of silica gel 60 features different grades with particle sizes ranging from 5-40 µm: silica with a gypsum binder, silica with no foreign binder, and silica gel with a fluorescence indicator. We also offer high-quality aluminum oxide, cellulose microcrystalline, and kieselgur (also known as diatomaceous earth).

Application


  • Chromatographic Separation of the Novel Hypoglycemic Drug Mitiglinide in its Bulk Powder and Pharmaceutical Formulation: Forced Degradation and Validated Stability-Indicating HPTLC/Densitometry and HPLC/UV Methods.: This research demonstrates the use of Silica gel 60 GF254 in the chromatographic separation of Mitiglinide, providing robust methods for stability and degradation analysis crucial for regulatory compliance and quality assurance in pharmaceuticals (Eissa MS et al., 2020).

  • Quality Risk Management-Based AQbD Approach to Development of VEER Chromatography Method for the Estimation of Multiple Combined Formulations of Anti-Hypertensive Drugs.: Leveraging Silica gel 60 GF254, this study outlines a novel chromatographic method development under the Analytical Quality by Design (AQbD) framework, enhancing the reliability and efficiency of drug analysis processes (Prajapati PB et al., 2021).

  • HPTLC and Spectrophotometric Estimation of Febuxostat and Diclofenac Potassium in Their Combined Tablets.: Utilizing Silica gel 60 GF254, this study presents a high-performance thin-layer chromatographic method for simultaneous estimation of Febuxostat and Diclofenac, demonstrating the gels utility in multi-drug analysis (El-Yazbi FA et al., 2016).

  • Distribution of withaferin A, an anticancer potential agent, in different parts of two varieties of Withania somnifera (L.) Dunal. grown in Sri Lanka.: This research used Silica gel 60 GF254 in the quantification of withaferin A across different plant parts, highlighting its application in the study of pharmacologically active compounds in medicinal plants (Siriwardane AS et al., 2013).

  • Simultaneous determination of metronidazole and miconazole nitrate in gel by HPTLC.: Demonstrating the adaptability of Silica gel 60 GF254, this study provides a validated HPTLC method for analyzing metronidazole and miconazole nitrate in a topical gel, crucial for ensuring therapeutic efficacy and safety in dermatological treatments (Meshram DB et al., 2009).

Analysis Note

pH-value (10% suspension): 6.0 - 7.0
Fe (Iron): ≤ 0.02%
Calcium sulfate (CaSO₄·0.5 H₂O): 12.0 - 13.5%
Particle size: 90% <55 µm
Application test: conforms
UV absorption (Dichloro methane extract): conforms
The data to the particle size are related to the pure Silica gel before additive addition.

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service