1.07733
Silica gel 60 (0.2-0.5 mm)
for column chromatography
Synonym(s):
Silica, Silicon dioxide
Select a Size
€58.00
Estimated to ship on30 May 2025
Select a Size
About This Item
€58.00
Estimated to ship on30 May 2025
Recommended Products
material
silica gel (irregularly-shaped)
Quality Level
matrix active group
Irregular-Shaped Silica Gel
particle size
200-500 μm
pore size
6 nm mean pore size
pH
7 (20 °C, 100 g/L in H2O, slurry)
bp
2230 °C (lit.)
mp
>1600 °C (lit.)
bulk density
200‑1430 kg/m3
application(s)
plasma purification
storage temp.
no temp limit
SMILES string
O=[Si]=O
InChI
1S/O2Si/c1-3-2
InChI key
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
Features and Benefits
- High adsorptive capacity
- High chemical inertness and non-toxicity.
- Protective moisture-sensitive substances during storage and transport or for maintaining the dryness of anhydrous solvents.
- Granulates in different sizes or beads
Packaging
Analysis Note
Fe (Iron): ≤ 0.02 %
Loss on drying: ≤ 7.0 %
Particle size: 90 % between 0.200 - 0.500 mm
Storage Class Code
13 - Non Combustible Solids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service