Skip to Content
Merck
All Photos(2)

Key Documents

1.07741

Supelco

Silica gel 60 HF₂₅₄+₃₆₆

for thin-layer chromatography

Synonym(s):

Silica gel

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41115711
EC Index Number:
231-545-4

material

plastic support
silica gel 60 matrix

Quality Level

form

powder

potency

>5000 mg/kg LD50, skin (Rabbit)

feature

binder: no
fluorescent indicator

packaging

pkg of 1 kg
bottle of

technique(s)

thin layer chromatography (TLC): suitable

particle size

5-40 μm

pore size

60 Å medium pore diameter

pH

7 (20 °C, 100 g/L in H2O, slurry)

bp

2230 °C

mp

>1600 °C

bulk density

200‑800 kg/m3

storage temp.

no temp limit

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

1 plastic bottle of 1 liter

Analysis Note

pH-value (10% suspension): 6.5 - 7.5
Fe (Iron): ≤ 0.02%
Particle size: 90% <55 µm
Application test: conforms
The data to the particle size are related to the pure Silica gel before additive addtition.

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service