Skip to Content
Merck

Skip To

1.04875

Potassium Phosphate Monobasic solution

(buffer stock solution) 1/15mol/l

Synonym(s):

Potassium Phosphate Monobasic solution, Potassium dihydrogen phosphate solution, Monopotassium phosphate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
1 L
Check Cart for Availability
€ 83,00

About This Item

CAS Number:
UNSPSC Code:
12161703
NACRES:
NA.24
MDL number:

€ 83,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Segment

density

1.005 g/cm3

application(s)

environmental
food and beverages
general analytical
industrial qc
pharmaceutical

format

mixture

storage temp.

15-25°C

SMILES string

[K+].OP(O)([O-])=O

InChI

1S/K.H3O4P/c;1-5(2,3)4/h;(H3,1,2,3,4)/q+1;/p-1

InChI key

GNSKLFRGEWLPPA-UHFFFAOYSA-M

General description

The accuracy of pH measurement results is directly affected by the buffer reference materials used in calibration. Merck offers you a distinguished range of buffer solutions and substances for the exact calibration of your pH measurement equipment. Using Certipur® reference materials, you benefit from maximum accuracy, reliability and convenience and avoid costly repeat analyses. For your strictest demands in equipment calibration and monitoring, we provide certified secondary standard buffer solutions and substances offering the highest precision.

Our wide range of pH calibration buffers offers ideal products and packaging for your individual need. All buffer solutions are certified reference materials according to ISO 17034 and qualified in our DIN EN ISO/IEC 17025 accredited laboratory. They are all traceable to NIST (National Institute of Standards and Technology, USA) and PTB (Physical Technical Institute, Germany) primary reference materials. Buffers solutions are offered in plastic bottles, measured at 20 °C or 25 °C.

Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation.

Download your certificate at http://www.sigma-aldrich.com to view certified values, including uncertainty and expiry date.

Application

Potassium dihydrogen phosphate, a monophosphate, is a certified reference buffer solution (CRM) used as a pH buffering agent, typically in combination with dipotassium phosphate.

Analysis Note

concentration: 0.0664 - 0.0670 mol/l

Legal Information

CERTIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
92214P87091551139
format

mixture

format

neat

format

-

format

neat

grade

certified reference material

grade

certified reference material, TraceCERT®

grade

reagent grade

grade

pharmaceutical primary standard

application(s)

environmental
food and beverages
general analytical
industrial qc
pharmaceutical

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care
pharmaceutical

application(s)

-

application(s)

pharmaceutical (small molecule)

storage temp.

15-25°C

storage temp.

-

storage temp.

-

storage temp.

-

density

1.005 g/cm3

density

2.338 g/mL at 25 °C (lit.)

density

-

density

2.338 g/mL at 25 °C (lit.)

Quality Level

100

Quality Level

100, 300

Quality Level

200

Quality Level

-


Still not finding the right product?


Storage Class

12 - Non Combustible Liquids

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library





Global Trade Item Number

SKUGTIN
104875100004022536056203

Questions

Reviews

No rating value

Active Filters